Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6638845-6639116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433967 c.1493G>T p.R498I Chr.12: 6638970-6638970 on Build GRCh38
1704984 c.1436C>A p.T479K Chr.12: 6639027-6639027 on Build GRCh38
3872295 c.1467C>T p.S489S Chr.12: 6638996-6638996 on Build GRCh38
4615094 c.1487delT p.L496fs*2 Chr.12: 6638976-6638976 on Build GRCh38
1363791 c.1426-1G>A p.? Chr.12: 6639038-6639038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138068731 Chr.12: 6639003-6639003 on Build GRCh38
rs990297037 Chr.12: 6638935-6638935 on Build GRCh38
rs745904260 Chr.12: 6638959-6638959 on Build GRCh38
rs373586314 Chr.12: 6639054-6639054 on Build GRCh38
rs923937497 Chr.12: 6639071-6639071 on Build GRCh38
rs936186006 Chr.12: 6638910-6638910 on Build GRCh38
rs765824351 Chr.12: 6639066-6639066 on Build GRCh38
rs774798304 Chr.12: 6638942-6638942 on Build GRCh38
rs377188946 Chr.12: 6639073-6639073 on Build GRCh38
rs779982883 Chr.12: 6638964-6638964 on Build GRCh38
rs772048700 Chr.12: 6638961-6638961 on Build GRCh38
rs12826388 Chr.12: 6638869-6638869 on Build GRCh38
rs754149345 Chr.12: 6639076-6639076 on Build GRCh38
rs577111362 Chr.12: 6638870-6638870 on Build GRCh38
rs1018778774 Chr.12: 6638922-6638922 on Build GRCh38
rs746905904 Chr.12: 6638965-6638965 on Build GRCh38
rs779662434 Chr.12: 6638928-6638930 on Build GRCh38
rs777988588 Chr.12: 6638909-6638909 on Build GRCh38
rs757645652 Chr.12: 6639083-6639083 on Build GRCh38
rs779126758 Chr.12: 6638958-6638958 on Build GRCh38
rs760123504 Chr.12: 6639063-6639064 on Build GRCh38
rs565391195 Chr.12: 6638890-6638890 on Build GRCh38
rs756447017 Chr.12: 6638907-6638907 on Build GRCh38
rs761084145 Chr.12: 6639067-6639067 on Build GRCh38
rs753398961 Chr.12: 6638876-6638876 on Build GRCh38
rs938998768 Chr.12: 6639075-6639075 on Build GRCh38
rs906182261 Chr.12: 6639061-6639061 on Build GRCh38
rs753153592 Chr.12: 6638903-6638903 on Build GRCh38
rs757446411 Chr.12: 6638919-6638919 on Build GRCh38
rs754239061 Chr.12: 6638917-6638917 on Build GRCh38
rs541437635 Chr.12: 6639044-6639044 on Build GRCh38
rs975219703 Chr.12: 6638888-6638888 on Build GRCh38
rs776537743 Chr.12: 6638981-6638981 on Build GRCh38
rs769522677 Chr.12: 6639017-6639017 on Build GRCh38
rs768703879 Chr.12: 6638975-6638975 on Build GRCh38
rs140069001 Chr.12: 6638872-6638872 on Build GRCh38
rs199953393 Chr.12: 6639020-6639020 on Build GRCh38
rs764631881 Chr.12: 6639069-6639069 on Build GRCh38

View Full Product Details