Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45402709-45402938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP27
Entrez Gene ID
201176
Gene Name
Rho GTPase activating protein 27
Gene Aliases
CAMGAP1, PP905, SH3D20, SH3P20
UniGene
Hs.205326
Gene Chromosome Location
Chr.17: 45393902-45432916 on Build GRCh38
Transcripts
NM_001282290.1 NM_199282.2 XM_005257128.3 XM_006721745.2 XM_006721747.2 XM_006721750.3 XM_011524468.1 XM_011524469.1 XM_011524470.2 XM_011524471.1 XM_011524472.2 XM_011524473.2 XM_011524474.1 XM_017024311.1 XM_017024312.1 XM_017024313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802954 c.688A>G p.K230E Chr.17: 45402746-45402746 on Build GRCh38
7356200 c.1655T>G p.F552C Chr.17: 45402802-45402802 on Build GRCh38
5091882 c.1699_1700delTC p.S567fs*9 Chr.17: 45402757-45402758 on Build GRCh38
4662826 c.656A>G p.E219G Chr.17: 45402778-45402778 on Build GRCh38
5013180 c.1704G>C p.W568C Chr.17: 45402753-45402753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17630935 Chr.17: 45402732-45402732 on Build GRCh38
rs925236204 Chr.17: 45402901-45402901 on Build GRCh38
rs750109008 Chr.17: 45402756-45402756 on Build GRCh38
rs979651434 Chr.17: 45402900-45402900 on Build GRCh38
rs773268491 Chr.17: 45402778-45402778 on Build GRCh38
rs143997699 Chr.17: 45402802-45402802 on Build GRCh38
rs774189135 Chr.17: 45402743-45402743 on Build GRCh38
rs368980069 Chr.17: 45402768-45402768 on Build GRCh38
rs751940489 Chr.17: 45402843-45402843 on Build GRCh38
rs767217091 Chr.17: 45402752-45402752 on Build GRCh38
rs375672160 Chr.17: 45402825-45402825 on Build GRCh38
rs762924863 Chr.17: 45402731-45402731 on Build GRCh38
rs922980735 Chr.17: 45402914-45402914 on Build GRCh38
rs56061413 Chr.17: 45402895-45402895 on Build GRCh38
rs966556952 Chr.17: 45402896-45402896 on Build GRCh38
rs1020365671 Chr.17: 45402880-45402880 on Build GRCh38
rs974438033 Chr.17: 45402913-45402913 on Build GRCh38
rs760227322 Chr.17: 45402786-45402786 on Build GRCh38
rs371691958 Chr.17: 45402906-45402906 on Build GRCh38
rs193141882 Chr.17: 45402870-45402870 on Build GRCh38
rs772209485 Chr.17: 45402750-45402750 on Build GRCh38
rs765938858 Chr.17: 45402789-45402789 on Build GRCh38
rs267604916 Chr.17: 45402799-45402799 on Build GRCh38
rs778211805 Chr.17: 45402827-45402827 on Build GRCh38
rs768512487 Chr.17: 45402734-45402734 on Build GRCh38

View Full Product Details