Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54592461-54592718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4577450 c.317G>T p.G106V Chr.1: 54592551-54592551 on Build GRCh38
4577451 c.317G>T p.G106V Chr.1: 54592551-54592551 on Build GRCh38
4008935 c.344T>C p.V115A Chr.1: 54592578-54592578 on Build GRCh38
1296585 c.370G>C p.E124Q Chr.1: 54592604-54592604 on Build GRCh38
5036822 c.315T>A p.V105V Chr.1: 54592549-54592549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986155770 Chr.1: 54592698-54592698 on Build GRCh38
rs183099244 Chr.1: 54592675-54592675 on Build GRCh38
rs758240522 Chr.1: 54592583-54592583 on Build GRCh38
rs572327660 Chr.1: 54592640-54592640 on Build GRCh38
rs757462544 Chr.1: 54592600-54592600 on Build GRCh38
rs1046781122 Chr.1: 54592660-54592660 on Build GRCh38
rs17394626 Chr.1: 54592487-54592487 on Build GRCh38
rs377614121 Chr.1: 54592507-54592507 on Build GRCh38
rs752752663 Chr.1: 54592582-54592582 on Build GRCh38
rs769075327 Chr.1: 54592506-54592506 on Build GRCh38
rs745902991 Chr.1: 54592607-54592607 on Build GRCh38
rs759118061 Chr.1: 54592552-54592552 on Build GRCh38
rs114817003 Chr.1: 54592500-54592500 on Build GRCh38
rs760423002 Chr.1: 54592514-54592514 on Build GRCh38
rs957342450 Chr.1: 54592532-54592532 on Build GRCh38
rs749831957 Chr.1: 54592625-54592625 on Build GRCh38
rs1030215422 Chr.1: 54592669-54592669 on Build GRCh38
rs955450690 Chr.1: 54592488-54592488 on Build GRCh38
rs886805137 Chr.1: 54592678-54592678 on Build GRCh38
rs376455629 Chr.1: 54592659-54592659 on Build GRCh38
rs998328051 Chr.1: 54592645-54592645 on Build GRCh38
rs373276988 Chr.1: 54592535-54592535 on Build GRCh38
rs370195766 Chr.1: 54592633-54592633 on Build GRCh38
rs765257577 Chr.1: 54592553-54592553 on Build GRCh38
rs990525336 Chr.1: 54592545-54592545 on Build GRCh38
rs1031392161 Chr.1: 54592598-54592598 on Build GRCh38
rs764002069 Chr.1: 54592584-54592584 on Build GRCh38
rs373473682 Chr.1: 54592641-54592641 on Build GRCh38
rs751335616 Chr.1: 54592593-54592593 on Build GRCh38
rs1005191389 Chr.1: 54592679-54592679 on Build GRCh38
rs756243665 Chr.1: 54592615-54592615 on Build GRCh38
rs576036843 Chr.1: 54592499-54592499 on Build GRCh38
rs947925334 Chr.1: 54592619-54592619 on Build GRCh38
rs192781341 Chr.1: 54592603-54592603 on Build GRCh38
rs369205658 Chr.1: 54592512-54592512 on Build GRCh38
rs774725619 Chr.1: 54592639-54592639 on Build GRCh38
rs780533901 Chr.1: 54592616-54592616 on Build GRCh38

View Full Product Details