Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94982949-94983204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG14
Entrez Gene ID
199857
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CMS15
UniGene
Hs.408927
Gene Chromosome Location
Chr.1: 94981565-95072974 on Build GRCh38
Transcripts
NM_001305242.1 NM_144988.3 NR_131032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1345036 c.641G>A p.R214Q Chr.1: 94983086-94983086 on Build GRCh38
5847521 c.626C>T p.S209L Chr.1: 94983101-94983101 on Build GRCh38
7355194 c.563T>C p.L188P Chr.1: 94983164-94983164 on Build GRCh38
6570788 c.562C>T p.L188L Chr.1: 94983165-94983165 on Build GRCh38
7210426 c.599C>T p.P200L Chr.1: 94983128-94983128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779018163 Chr.1: 94983111-94983111 on Build GRCh38
rs113133699 Chr.1: 94983076-94983076 on Build GRCh38
rs955691169 Chr.1: 94982988-94982988 on Build GRCh38
rs767036165 Chr.1: 94983086-94983086 on Build GRCh38
rs760367713 Chr.1: 94983069-94983069 on Build GRCh38
rs774952403 Chr.1: 94983179-94983179 on Build GRCh38
rs979844768 Chr.1: 94983013-94983013 on Build GRCh38
rs949825162 Chr.1: 94983006-94983006 on Build GRCh38
rs754944986 Chr.1: 94983101-94983101 on Build GRCh38
rs1040766612 Chr.1: 94983068-94983068 on Build GRCh38
rs750095172 Chr.1: 94983087-94983087 on Build GRCh38
rs112335700 Chr.1: 94983069-94983069 on Build GRCh38
rs755627570 Chr.1: 94983089-94983089 on Build GRCh38
rs1050499608 Chr.1: 94983094-94983094 on Build GRCh38
rs913400574 Chr.1: 94982999-94982999 on Build GRCh38
rs200959923 Chr.1: 94983118-94983118 on Build GRCh38
rs946171388 Chr.1: 94983008-94983008 on Build GRCh38
rs200609853 Chr.1: 94983122-94983122 on Build GRCh38
rs570453160 Chr.1: 94982982-94982982 on Build GRCh38
rs764623435 Chr.1: 94983059-94983059 on Build GRCh38
rs3198368 Chr.1: 94983116-94983116 on Build GRCh38
rs148202016 Chr.1: 94983175-94983175 on Build GRCh38
rs1014644772 Chr.1: 94983128-94983128 on Build GRCh38
rs765319440 Chr.1: 94983037-94983037 on Build GRCh38
rs995053145 Chr.1: 94983150-94983150 on Build GRCh38
rs143040411 Chr.1: 94983063-94983063 on Build GRCh38
rs770065162 Chr.1: 94983157-94983157 on Build GRCh38
rs547996899 Chr.1: 94983005-94983006 on Build GRCh38
rs759471629 Chr.1: 94983029-94983029 on Build GRCh38
rs746084809 Chr.1: 94983144-94983144 on Build GRCh38
rs763442364 Chr.1: 94983055-94983055 on Build GRCh38
rs901868737 Chr.1: 94983097-94983097 on Build GRCh38
rs552681984 Chr.1: 94983020-94983020 on Build GRCh38
rs757661309 Chr.1: 94983064-94983064 on Build GRCh38
rs776663396 Chr.1: 94983129-94983129 on Build GRCh38
rs775514355 Chr.1: 94983167-94983167 on Build GRCh38
rs555245521 Chr.1: 94983091-94983091 on Build GRCh38
rs961883242 Chr.1: 94983148-94983148 on Build GRCh38
rs539550497 Chr.1: 94983004-94983004 on Build GRCh38
rs770387900 Chr.1: 94983100-94983100 on Build GRCh38
rs996262392 Chr.1: 94983099-94983099 on Build GRCh38
rs886374685 Chr.1: 94983050-94983050 on Build GRCh38
rs775600527 Chr.1: 94983041-94983041 on Build GRCh38
rs771908149 Chr.1: 94983127-94983127 on Build GRCh38
rs199846785 Chr.1: 94983181-94983181 on Build GRCh38
rs777079109 Chr.1: 94983033-94983033 on Build GRCh38
rs376803101 Chr.1: 94983031-94983031 on Build GRCh38
rs916976401 Chr.1: 94983006-94983006 on Build GRCh38
rs79206206 Chr.1: 94983022-94983022 on Build GRCh38
rs764533791 Chr.1: 94983178-94983178 on Build GRCh38

View Full Product Details