Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31086319-31086578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCYAP1R1
Entrez Gene ID
117
Gene Name
ADCYAP receptor type I
Gene Aliases
PAC1, PAC1R, PACAPR, PACAPRI
UniGene
Hs.377783
Gene Chromosome Location
Chr.7: 31052341-31114005 on Build GRCh38
Transcripts
NM_001118.4 NM_001199635.1 NM_001199636.1 NM_001199637.1 XM_005249618.4 XM_006715645.2 XM_017011736.1 XM_017011737.1 XM_017011738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110055 c.680A>T p.K227M Chr.7: 31086394-31086394 on Build GRCh38
3637981 c.800A>G p.Y267C Chr.7: 31086514-31086514 on Build GRCh38
1161675 c.730C>T p.L244L Chr.7: 31086444-31086444 on Build GRCh38
6595086 c.781C>A p.P261T Chr.7: 31086495-31086495 on Build GRCh38
7255169 c.735C>T p.F245F Chr.7: 31086449-31086449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549741398 Chr.7: 31086369-31086369 on Build GRCh38
rs753089157 Chr.7: 31086556-31086599 on Build GRCh38
rs150564353 Chr.7: 31086452-31086452 on Build GRCh38
rs1036947393 Chr.7: 31086384-31086384 on Build GRCh38
rs757943758 Chr.7: 31086524-31086524 on Build GRCh38
rs753778697 Chr.7: 31086341-31086341 on Build GRCh38
rs751267185 Chr.7: 31086539-31086539 on Build GRCh38
rs993479822 Chr.7: 31086422-31086422 on Build GRCh38
rs552956438 Chr.7: 31086468-31086468 on Build GRCh38
rs778377419 Chr.7: 31086361-31086362 on Build GRCh38
rs199506110 Chr.7: 31086401-31086401 on Build GRCh38
rs372962736 Chr.7: 31086358-31086358 on Build GRCh38
rs756944794 Chr.7: 31086552-31086552 on Build GRCh38
rs561323893 Chr.7: 31086416-31086416 on Build GRCh38
rs777256204 Chr.7: 31086537-31086537 on Build GRCh38
rs890470937 Chr.7: 31086356-31086356 on Build GRCh38
rs769239511 Chr.7: 31086406-31086406 on Build GRCh38
rs766455986 Chr.7: 31086456-31086456 on Build GRCh38
rs903340336 Chr.7: 31086559-31086559 on Build GRCh38
rs138384179 Chr.7: 31086355-31086355 on Build GRCh38
rs772251803 Chr.7: 31086421-31086421 on Build GRCh38
rs1044247186 Chr.7: 31086347-31086347 on Build GRCh38
rs749555552 Chr.7: 31086402-31086402 on Build GRCh38
rs759210023 Chr.7: 31086473-31086473 on Build GRCh38
rs746633319 Chr.7: 31086398-31086398 on Build GRCh38
rs759125342 Chr.7: 31086474-31086474 on Build GRCh38
rs963457938 Chr.7: 31086455-31086455 on Build GRCh38
rs375760330 Chr.7: 31086354-31086354 on Build GRCh38
rs762036945 Chr.7: 31086418-31086418 on Build GRCh38
rs757853879 Chr.7: 31086374-31086374 on Build GRCh38
rs866491053 Chr.7: 31086432-31086432 on Build GRCh38
rs765240963 Chr.7: 31086482-31086482 on Build GRCh38
rs753673499 Chr.7: 31086461-31086461 on Build GRCh38
rs770206447 Chr.7: 31086399-31086399 on Build GRCh38
rs777449969 Chr.7: 31086378-31086378 on Build GRCh38
rs372588011 Chr.7: 31086346-31086346 on Build GRCh38
rs141519354 Chr.7: 31086494-31086494 on Build GRCh38

View Full Product Details