Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50696895-50697166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD40
Entrez Gene ID
91369
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
UniGene
Hs.463426
Gene Chromosome Location
Chr.17: 50693190-50707909 on Build GRCh38
Transcripts
NM_052855.3 XM_017025296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661889 c.784G>T p.V262L Chr.17: 50697116-50697116 on Build GRCh38
4661888 c.797G>T p.R266I Chr.17: 50697103-50697103 on Build GRCh38
1384367 c.850C>T p.R284* Chr.17: 50697050-50697050 on Build GRCh38
3819972 c.794T>G p.V265G Chr.17: 50697106-50697106 on Build GRCh38
5774602 c.833T>C p.I278T Chr.17: 50697067-50697067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762144895 Chr.17: 50697125-50697126 on Build GRCh38
rs757615177 Chr.17: 50696997-50696997 on Build GRCh38
rs775077880 Chr.17: 50697128-50697128 on Build GRCh38
rs747530576 Chr.17: 50697032-50697032 on Build GRCh38
rs184264254 Chr.17: 50697135-50697135 on Build GRCh38
rs563704829 Chr.17: 50696948-50696948 on Build GRCh38
rs548025944 Chr.17: 50697039-50697039 on Build GRCh38
rs777053408 Chr.17: 50697023-50697024 on Build GRCh38
rs772776762 Chr.17: 50697050-50697050 on Build GRCh38
rs765478207 Chr.17: 50697126-50697126 on Build GRCh38
rs764635009 Chr.17: 50696939-50696939 on Build GRCh38
rs758825406 Chr.17: 50697007-50697007 on Build GRCh38
rs149520387 Chr.17: 50697001-50697001 on Build GRCh38
rs756055314 Chr.17: 50696965-50696965 on Build GRCh38
rs3815378 Chr.17: 50696958-50696958 on Build GRCh38
rs780234518 Chr.17: 50697009-50697009 on Build GRCh38
rs201524009 Chr.17: 50697125-50697125 on Build GRCh38
rs770610835 Chr.17: 50697082-50697082 on Build GRCh38
rs750685969 Chr.17: 50697133-50697134 on Build GRCh38
rs753958189 Chr.17: 50696993-50696993 on Build GRCh38
rs774082706 Chr.17: 50697096-50697096 on Build GRCh38
rs926266240 Chr.17: 50696949-50696949 on Build GRCh38
rs759060373 Chr.17: 50697097-50697097 on Build GRCh38
rs764270751 Chr.17: 50696980-50696980 on Build GRCh38
rs748773517 Chr.17: 50697051-50697051 on Build GRCh38
rs779178026 Chr.17: 50696999-50696999 on Build GRCh38
rs767354676 Chr.17: 50697116-50697116 on Build GRCh38
rs144124178 Chr.17: 50697017-50697017 on Build GRCh38
rs769085636 Chr.17: 50697041-50697041 on Build GRCh38

View Full Product Details