Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81711284-81711535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3882610 c.1427C>T p.T476I Chr.7: 81711498-81711498 on Build GRCh38
3030144 c.1444C>A p.H482N Chr.7: 81711481-81711481 on Build GRCh38
6971407 c.1444+1G>C p.? Chr.7: 81711480-81711480 on Build GRCh38
7130882 c.1441G>T p.D481Y Chr.7: 81711484-81711484 on Build GRCh38
6925975 c.1444+1G>A p.? Chr.7: 81711480-81711480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185526560 Chr.7: 81711456-81711456 on Build GRCh38
rs573915342 Chr.7: 81711424-81711424 on Build GRCh38
rs370419243 Chr.7: 81711474-81711474 on Build GRCh38
rs769943357 Chr.7: 81711433-81711433 on Build GRCh38
rs5745736 Chr.7: 81711425-81711425 on Build GRCh38
rs761396857 Chr.7: 81711481-81711481 on Build GRCh38
rs763802488 Chr.7: 81711475-81711475 on Build GRCh38
rs764922029 Chr.7: 81711503-81711503 on Build GRCh38
rs986458402 Chr.7: 81711418-81711418 on Build GRCh38
rs568418022 Chr.7: 81711398-81711398 on Build GRCh38
rs925158097 Chr.7: 81711380-81711380 on Build GRCh38
rs936053876 Chr.7: 81711412-81711412 on Build GRCh38
rs560462747 Chr.7: 81711399-81711399 on Build GRCh38
rs980560459 Chr.7: 81711346-81711346 on Build GRCh38
rs767889 Chr.7: 81711429-81711429 on Build GRCh38
rs564721078 Chr.7: 81711480-81711480 on Build GRCh38
rs969301779 Chr.7: 81711323-81711323 on Build GRCh38
rs562185059 Chr.7: 81711435-81711435 on Build GRCh38
rs772131129 Chr.7: 81711450-81711450 on Build GRCh38
rs763164743 Chr.7: 81711443-81711443 on Build GRCh38

View Full Product Details