Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231265583-231265844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPAT
Entrez Gene ID
8443
Gene Name
glyceronephosphate O-acyltransferase
Gene Aliases
DAP-AT, DAPAT, DHAPAT, RCDP2
UniGene
Hs.498028
Gene Chromosome Location
Chr.1: 231241173-231277973 on Build GRCh38
Transcripts
NM_001316350.1 NM_014236.3 XM_005273313.4 XM_011544303.2 XM_011544304.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1178041 c.747T>A p.S249S Chr.1: 231265762-231265762 on Build GRCh38
4506551 c.723C>T p.F241F Chr.1: 231265738-231265738 on Build GRCh38
905466 c.721T>G p.F241V Chr.1: 231265736-231265736 on Build GRCh38
3400460 c.742C>T p.R248C Chr.1: 231265757-231265757 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759286038 Chr.1: 231265710-231265710 on Build GRCh38
rs771696243 Chr.1: 231265772-231265772 on Build GRCh38
rs374437334 Chr.1: 231265713-231265713 on Build GRCh38
rs776581840 Chr.1: 231265798-231265798 on Build GRCh38
rs777756556 Chr.1: 231265790-231265790 on Build GRCh38
rs755504902 Chr.1: 231265727-231265727 on Build GRCh38
rs778571132 Chr.1: 231265753-231265753 on Build GRCh38
rs753865102 Chr.1: 231265693-231265693 on Build GRCh38
rs756746406 Chr.1: 231265752-231265752 on Build GRCh38
rs746749312 Chr.1: 231265783-231265783 on Build GRCh38
rs780614492 Chr.1: 231265724-231265724 on Build GRCh38
rs375170975 Chr.1: 231265781-231265781 on Build GRCh38
rs545225435 Chr.1: 231265661-231265661 on Build GRCh38
rs781676879 Chr.1: 231265739-231265739 on Build GRCh38
rs765197038 Chr.1: 231265773-231265773 on Build GRCh38
rs775239940 Chr.1: 231265667-231265667 on Build GRCh38
rs758663178 Chr.1: 231265721-231265721 on Build GRCh38
rs776313541 Chr.1: 231265658-231265658 on Build GRCh38
rs760583712 Chr.1: 231265677-231265677 on Build GRCh38
rs750632636 Chr.1: 231265720-231265720 on Build GRCh38
rs565061169 Chr.1: 231265812-231265812 on Build GRCh38
rs202033127 Chr.1: 231265762-231265762 on Build GRCh38
rs761840137 Chr.1: 231265800-231265800 on Build GRCh38
rs764085916 Chr.1: 231265681-231265681 on Build GRCh38
rs145974748 Chr.1: 231265715-231265715 on Build GRCh38
rs909786991 Chr.1: 231265630-231265630 on Build GRCh38
rs748763962 Chr.1: 231265742-231265742 on Build GRCh38

View Full Product Details