Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46031928-46032185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
706071 c.3012T>C p.P1004P Chr.17: 46032125-46032125 on Build GRCh38
980236 c.3052A>G p.T1018A Chr.17: 46032085-46032085 on Build GRCh38
4485611 c.2952C>T p.S984S Chr.17: 46032185-46032185 on Build GRCh38
5629830 c.3057T>G p.R1019R Chr.17: 46032080-46032080 on Build GRCh38
4915006 c.3089A>G p.Q1030R Chr.17: 46032048-46032048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895906847 Chr.17: 46031967-46031967 on Build GRCh38
rs748843892 Chr.17: 46032107-46032107 on Build GRCh38
rs373179809 Chr.17: 46031956-46031956 on Build GRCh38
rs752833970 Chr.17: 46032064-46032064 on Build GRCh38
rs745540054 Chr.17: 46031999-46031999 on Build GRCh38
rs781056926 Chr.17: 46032081-46032081 on Build GRCh38
rs371484505 Chr.17: 46032152-46032152 on Build GRCh38
rs758333951 Chr.17: 46032067-46032067 on Build GRCh38
rs748649772 Chr.17: 46032008-46032008 on Build GRCh38
rs765297359 Chr.17: 46032055-46032055 on Build GRCh38
rs779686052 Chr.17: 46032106-46032106 on Build GRCh38
rs946207471 Chr.17: 46031983-46031983 on Build GRCh38
rs769373626 Chr.17: 46032000-46032000 on Build GRCh38
rs772500977 Chr.17: 46032012-46032012 on Build GRCh38
rs780672258 Chr.17: 46031998-46031998 on Build GRCh38
rs145863194 Chr.17: 46032084-46032084 on Build GRCh38
rs776773399 Chr.17: 46032026-46032026 on Build GRCh38
rs7220988 Chr.17: 46032108-46032108 on Build GRCh38
rs1037585739 Chr.17: 46032135-46032135 on Build GRCh38
rs201824489 Chr.17: 46032004-46032004 on Build GRCh38
rs751551522 Chr.17: 46032072-46032072 on Build GRCh38
rs974482953 Chr.17: 46032035-46032035 on Build GRCh38
rs775516828 Chr.17: 46032129-46032129 on Build GRCh38
rs376138960 Chr.17: 46032017-46032017 on Build GRCh38
rs773591156 Chr.17: 46032015-46032015 on Build GRCh38
rs138490347 Chr.17: 46032099-46032099 on Build GRCh38
rs573432386 Chr.17: 46032126-46032126 on Build GRCh38
rs148203818 Chr.17: 46032117-46032117 on Build GRCh38
rs761819132 Chr.17: 46032153-46032153 on Build GRCh38
rs762905143 Chr.17: 46032134-46032134 on Build GRCh38
rs747564871 Chr.17: 46032110-46032110 on Build GRCh38
rs143018360 Chr.17: 46032109-46032109 on Build GRCh38
rs777045712 Chr.17: 46032112-46032112 on Build GRCh38
rs376488097 Chr.17: 46031997-46031997 on Build GRCh38
rs764260806 Chr.17: 46032146-46032146 on Build GRCh38
rs756958045 Chr.17: 46032075-46032075 on Build GRCh38
rs201117362 Chr.17: 46032038-46032038 on Build GRCh38
rs763978952 Chr.17: 46032071-46032071 on Build GRCh38

View Full Product Details