Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155240679-155240940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747794 c.45G>C p.L15F Chr.1: 155240700-155240700 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769807351 Chr.1: 155240759-155240759 on Build GRCh38
rs2070679 Chr.1: 155240850-155240850 on Build GRCh38
rs778920100 Chr.1: 155240727-155240727 on Build GRCh38
rs755682174 Chr.1: 155240709-155240709 on Build GRCh38
rs150466109 Chr.1: 155240707-155240707 on Build GRCh38
rs1141802 Chr.1: 155240701-155240701 on Build GRCh38
rs745734072 Chr.1: 155240729-155240729 on Build GRCh38
rs749537308 Chr.1: 155240769-155240769 on Build GRCh38
rs543787905 Chr.1: 155240786-155240786 on Build GRCh38
rs372173221 Chr.1: 155240768-155240768 on Build GRCh38
rs758423368 Chr.1: 155240738-155240738 on Build GRCh38
rs2885305 Chr.1: 155240752-155240752 on Build GRCh38
rs1045935620 Chr.1: 155240797-155240797 on Build GRCh38
rs763770350 Chr.1: 155240711-155240711 on Build GRCh38
rs371238435 Chr.1: 155240705-155240705 on Build GRCh38
rs978014736 Chr.1: 155240718-155240718 on Build GRCh38
rs1037310264 Chr.1: 155240835-155240835 on Build GRCh38
rs774833277 Chr.1: 155240702-155240702 on Build GRCh38
rs2361534 Chr.1: 155240779-155240779 on Build GRCh38
rs940168433 Chr.1: 155240816-155240816 on Build GRCh38
rs571598853 Chr.1: 155240867-155240867 on Build GRCh38
rs748294651 Chr.1: 155240754-155240754 on Build GRCh38
rs753480979 Chr.1: 155240723-155240723 on Build GRCh38
rs528452955 Chr.1: 155240803-155240803 on Build GRCh38
rs757059817 Chr.1: 155240725-155240725 on Build GRCh38
rs868719358 Chr.1: 155240765-155240765 on Build GRCh38
rs773464365 Chr.1: 155240767-155240767 on Build GRCh38
rs2361535 Chr.1: 155240815-155240815 on Build GRCh38

View Full Product Details