Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154983807-154984080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLAD1
Entrez Gene ID
80308
Gene Name
flavin adenine dinucleotide synthetase 1
Gene Aliases
FAD1, FADS, LSMFLAD, PP591
UniGene
Hs.118666
Gene Chromosome Location
Chr.1: 154983294-154993111 on Build GRCh38
Transcripts
NM_001184891.1 NM_001184892.1 NM_025207.4 NM_201398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158446 c.238G>C p.G80R Chr.1: 154983932-154983932 on Build GRCh38
6664389 c.204C>T p.D68D Chr.1: 154983898-154983898 on Build GRCh38
4684810 c.116C>T p.A39V Chr.1: 154983810-154983810 on Build GRCh38
6564996 c.227G>A p.R76Q Chr.1: 154983921-154983921 on Build GRCh38
3802052 c.222C>A p.C74* Chr.1: 154983916-154983916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768686282 Chr.1: 154983851-154983851 on Build GRCh38
rs776207931 Chr.1: 154983901-154983901 on Build GRCh38
rs763542723 Chr.1: 154984032-154984032 on Build GRCh38
rs778858864 Chr.1: 154983971-154983971 on Build GRCh38
rs943456545 Chr.1: 154983939-154983939 on Build GRCh38
rs771819089 Chr.1: 154983881-154983881 on Build GRCh38
rs965450382 Chr.1: 154983911-154983911 on Build GRCh38
rs540907370 Chr.1: 154983935-154983935 on Build GRCh38
rs750944765 Chr.1: 154983910-154983910 on Build GRCh38
rs764492355 Chr.1: 154983974-154983981 on Build GRCh38
rs768383309 Chr.1: 154984007-154984007 on Build GRCh38
rs1035764473 Chr.1: 154983874-154983874 on Build GRCh38
rs150060495 Chr.1: 154983897-154983897 on Build GRCh38
rs559495946 Chr.1: 154984013-154984013 on Build GRCh38
rs756598623 Chr.1: 154984041-154984041 on Build GRCh38
rs780188906 Chr.1: 154983839-154983839 on Build GRCh38
rs908874898 Chr.1: 154983873-154983873 on Build GRCh38
rs775399217 Chr.1: 154983907-154983907 on Build GRCh38
rs775069508 Chr.1: 154983882-154983882 on Build GRCh38
rs770864376 Chr.1: 154983860-154983860 on Build GRCh38
rs774147030 Chr.1: 154983886-154983912 on Build GRCh38
rs150906684 Chr.1: 154983846-154983846 on Build GRCh38
rs876661314 Chr.1: 154984018-154984018 on Build GRCh38
rs756141523 Chr.1: 154983969-154983969 on Build GRCh38
rs375758484 Chr.1: 154984024-154984024 on Build GRCh38
rs767806305 Chr.1: 154984057-154984057 on Build GRCh38
rs773925274 Chr.1: 154984014-154984014 on Build GRCh38
rs1054257 Chr.1: 154984004-154984004 on Build GRCh38
rs760359719 Chr.1: 154983892-154983892 on Build GRCh38
rs748170567 Chr.1: 154983857-154983857 on Build GRCh38
rs544864359 Chr.1: 154983903-154983904 on Build GRCh38
rs561807633 Chr.1: 154983903-154983903 on Build GRCh38
rs759372173 Chr.1: 154983879-154983879 on Build GRCh38
rs781322149 Chr.1: 154983852-154983852 on Build GRCh38
rs941684349 Chr.1: 154983890-154983890 on Build GRCh38
rs775365908 Chr.1: 154984000-154984000 on Build GRCh38
rs766763725 Chr.1: 154983929-154983929 on Build GRCh38
rs770645984 Chr.1: 154983884-154983885 on Build GRCh38
rs368689507 Chr.1: 154984002-154984002 on Build GRCh38
rs1002583534 Chr.1: 154984046-154984046 on Build GRCh38
rs760857353 Chr.1: 154983940-154983940 on Build GRCh38
rs748262474 Chr.1: 154983965-154983965 on Build GRCh38
rs751960963 Chr.1: 154984044-154984044 on Build GRCh38
rs771817662 Chr.1: 154983982-154983982 on Build GRCh38
rs772370005 Chr.1: 154983906-154983907 on Build GRCh38
rs751787898 Chr.1: 154983932-154983932 on Build GRCh38
rs200341516 Chr.1: 154983926-154983926 on Build GRCh38
rs1044204334 Chr.1: 154984033-154984033 on Build GRCh38
rs745796104 Chr.1: 154983976-154983976 on Build GRCh38
rs781206370 Chr.1: 154983944-154983944 on Build GRCh38
rs749079149 Chr.1: 154983836-154983837 on Build GRCh38

View Full Product Details