Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32769620-32769891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1522
Entrez Gene ID
57648
Gene Name
KIAA1522
Gene Aliases
-
UniGene
Hs.591502
Gene Chromosome Location
Chr.1: 32741839-32774970 on Build GRCh38
Transcripts
NM_001198972.1 NM_001198973.1 NM_020888.2 XM_006710791.3 XM_017001916.1 XM_017001917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5163477 c.467G>C p.R156P Chr.1: 32769730-32769730 on Build GRCh38
7150469 c.655C>G p.R219G Chr.1: 32769741-32769741 on Build GRCh38
4621413 c.438T>C p.F146F Chr.1: 32769701-32769701 on Build GRCh38
6392907 c.652C>T p.R218W Chr.1: 32769738-32769738 on Build GRCh38
5778463 c.430G>C p.D144H Chr.1: 32769693-32769693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765214719 Chr.1: 32769689-32769689 on Build GRCh38
rs1054867809 Chr.1: 32769685-32769685 on Build GRCh38
rs769617710 Chr.1: 32769808-32769808 on Build GRCh38
rs780681784 Chr.1: 32769858-32769858 on Build GRCh38
rs372392689 Chr.1: 32769849-32769849 on Build GRCh38
rs759041305 Chr.1: 32769656-32769656 on Build GRCh38
rs779521825 Chr.1: 32769844-32769845 on Build GRCh38
rs754666751 Chr.1: 32769660-32769660 on Build GRCh38
rs776390424 Chr.1: 32769794-32769794 on Build GRCh38
rs372431951 Chr.1: 32769764-32769764 on Build GRCh38
rs927865462 Chr.1: 32769666-32769666 on Build GRCh38
rs755051799 Chr.1: 32769698-32769698 on Build GRCh38
rs765496689 Chr.1: 32769847-32769847 on Build GRCh38
rs760474797 Chr.1: 32769665-32769665 on Build GRCh38
rs761728481 Chr.1: 32769687-32769687 on Build GRCh38
rs753616662 Chr.1: 32769673-32769673 on Build GRCh38
rs762812559 Chr.1: 32769817-32769817 on Build GRCh38
rs199581538 Chr.1: 32769813-32769813 on Build GRCh38
rs184355507 Chr.1: 32769668-32769668 on Build GRCh38
rs746406963 Chr.1: 32769727-32769727 on Build GRCh38
rs749669036 Chr.1: 32769645-32769645 on Build GRCh38
rs982509436 Chr.1: 32769807-32769807 on Build GRCh38
rs531799490 Chr.1: 32769777-32769777 on Build GRCh38
rs959571742 Chr.1: 32769824-32769824 on Build GRCh38
rs758810870 Chr.1: 32769855-32769855 on Build GRCh38
rs372939895 Chr.1: 32769742-32769742 on Build GRCh38
rs377257501 Chr.1: 32769748-32769748 on Build GRCh38
rs561655625 Chr.1: 32769733-32769733 on Build GRCh38
rs367698882 Chr.1: 32769701-32769701 on Build GRCh38
rs761677287 Chr.1: 32769740-32769740 on Build GRCh38
rs527729808 Chr.1: 32769724-32769724 on Build GRCh38
rs74067513 Chr.1: 32769730-32769730 on Build GRCh38
rs771156476 Chr.1: 32769860-32769860 on Build GRCh38
rs764253546 Chr.1: 32769753-32769753 on Build GRCh38
rs750375738 Chr.1: 32769693-32769693 on Build GRCh38
rs760725071 Chr.1: 32769837-32769837 on Build GRCh38
rs766186539 Chr.1: 32769835-32769835 on Build GRCh38
rs754112149 Chr.1: 32769757-32769757 on Build GRCh38
rs779455146 Chr.1: 32769726-32769726 on Build GRCh38
rs756272278 Chr.1: 32769722-32769722 on Build GRCh38
rs752949839 Chr.1: 32769716-32769716 on Build GRCh38
rs776441775 Chr.1: 32769738-32769738 on Build GRCh38
rs760262368 Chr.1: 32769732-32769732 on Build GRCh38
rs1011697140 Chr.1: 32769712-32769712 on Build GRCh38
rs771611424 Chr.1: 32769729-32769729 on Build GRCh38
rs757809300 Chr.1: 32769764-32769766 on Build GRCh38
rs752707295 Chr.1: 32769745-32769745 on Build GRCh38
rs575829282 Chr.1: 32769845-32769845 on Build GRCh38
rs771361249 Chr.1: 32769655-32769655 on Build GRCh38
rs369365755 Chr.1: 32769789-32769789 on Build GRCh38
rs764978928 Chr.1: 32769741-32769741 on Build GRCh38
rs201274811 Chr.1: 32769763-32769763 on Build GRCh38
rs747558096 Chr.1: 32769859-32769859 on Build GRCh38
rs202166058 Chr.1: 32769770-32769770 on Build GRCh38
rs746377198 Chr.1: 32769766-32769766 on Build GRCh38
rs201768529 Chr.1: 32769838-32769838 on Build GRCh38
rs749711613 Chr.1: 32769655-32769665 on Build GRCh38
rs777988456 Chr.1: 32769644-32769645 on Build GRCh38
rs771533470 Chr.1: 32769659-32769659 on Build GRCh38
rs373341160 Chr.1: 32769805-32769805 on Build GRCh38
rs770139821 Chr.1: 32769759-32769759 on Build GRCh38
rs754059269 Chr.1: 32769842-32769842 on Build GRCh38
rs754528931 Chr.1: 32769863-32769863 on Build GRCh38
rs757590016 Chr.1: 32769758-32769758 on Build GRCh38
rs746466702 Chr.1: 32769778-32769778 on Build GRCh38

View Full Product Details