Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12942152-12942425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234B
Entrez Gene ID
57613
Gene Name
family with sequence similarity 234 member B
Gene Aliases
KIAA1467
UniGene
Hs.132660
Gene Chromosome Location
Chr.12: 12927680-13083449 on Build GRCh38
Transcripts
XM_017019706.1 NM_018654.1 XM_005253421.3 XM_017019582.1 XM_017019583.1

Gene Symbol
GPRC5D
Entrez Gene ID
55507
Gene Name
G protein-coupled receptor class C group 5 member D
Gene Aliases
-
UniGene
Hs.644599
Gene Chromosome Location
Chr.12: 12940518-12955908 on Build GRCh38
Transcripts
XM_017019706.1 NM_018654.1 XM_005253421.3 XM_017019582.1 XM_017019583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4446146 c.896-3C>T p.? Chr.12: 12942331-12942331 on Build GRCh38
2228439 c.947C>T p.T316I Chr.12: 12942277-12942277 on Build GRCh38
123451 c.943G>T p.G315C Chr.12: 12942281-12942281 on Build GRCh38
4980456 c.941A>C p.Y314S Chr.12: 12942283-12942283 on Build GRCh38
4840795 c.928G>A p.A310T Chr.12: 12942296-12942296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11055164 Chr.12: 12942310-12942310 on Build GRCh38
rs745554764 Chr.12: 12942245-12942245 on Build GRCh38
rs768574348 Chr.12: 12942318-12942318 on Build GRCh38
rs755354026 Chr.12: 12942267-12942267 on Build GRCh38
rs935807099 Chr.12: 12942250-12942250 on Build GRCh38
rs942410855 Chr.12: 12942195-12942195 on Build GRCh38
rs776056439 Chr.12: 12942234-12942234 on Build GRCh38
rs372223484 Chr.12: 12942336-12942336 on Build GRCh38
rs749637727 Chr.12: 12942217-12942217 on Build GRCh38
rs773185845 Chr.12: 12942379-12942379 on Build GRCh38
rs1017774996 Chr.12: 12942389-12942389 on Build GRCh38
rs1013670974 Chr.12: 12942381-12942381 on Build GRCh38
rs975106884 Chr.12: 12942196-12942196 on Build GRCh38
rs761773478 Chr.12: 12942261-12942261 on Build GRCh38
rs758953290 Chr.12: 12942349-12942349 on Build GRCh38
rs755638580 Chr.12: 12942355-12942355 on Build GRCh38
rs778221606 Chr.12: 12942351-12942351 on Build GRCh38
rs368934325 Chr.12: 12942197-12942197 on Build GRCh38
rs748661173 Chr.12: 12942277-12942277 on Build GRCh38
rs373908218 Chr.12: 12942264-12942264 on Build GRCh38
rs779194611 Chr.12: 12942271-12942271 on Build GRCh38
rs200107371 Chr.12: 12942369-12942369 on Build GRCh38
rs760802306 Chr.12: 12942327-12942327 on Build GRCh38
rs148322311 Chr.12: 12942280-12942280 on Build GRCh38
rs762779703 Chr.12: 12942251-12942251 on Build GRCh38
rs1047987711 Chr.12: 12942406-12942406 on Build GRCh38
rs778899601 Chr.12: 12942374-12942374 on Build GRCh38
rs373712100 Chr.12: 12942248-12942248 on Build GRCh38
rs1026954934 Chr.12: 12942367-12942367 on Build GRCh38
rs746582469 Chr.12: 12942230-12942230 on Build GRCh38
rs369852260 Chr.12: 12942229-12942229 on Build GRCh38
rs775198322 Chr.12: 12942249-12942249 on Build GRCh38
rs193015651 Chr.12: 12942354-12942354 on Build GRCh38
rs764005587 Chr.12: 12942255-12942255 on Build GRCh38
rs779362925 Chr.12: 12942365-12942365 on Build GRCh38
rs749492699 Chr.12: 12942361-12942361 on Build GRCh38
rs775205966 Chr.12: 12942306-12942306 on Build GRCh38
rs766564946 Chr.12: 12942329-12942329 on Build GRCh38
rs1041989971 Chr.12: 12942237-12942237 on Build GRCh38
rs372404743 Chr.12: 12942239-12942239 on Build GRCh38
rs186556261 Chr.12: 12942377-12942377 on Build GRCh38
rs752220590 Chr.12: 12942251-12942251 on Build GRCh38
rs202085045 Chr.12: 12942265-12942265 on Build GRCh38
rs201784839 Chr.12: 12942325-12942325 on Build GRCh38
rs755475721 Chr.12: 12942218-12942218 on Build GRCh38
rs201184166 Chr.12: 12942323-12942323 on Build GRCh38
rs756213833 Chr.12: 12942180-12942180 on Build GRCh38
rs749096153 Chr.12: 12942309-12942309 on Build GRCh38
rs373699086 Chr.12: 12942331-12942331 on Build GRCh38
rs370808293 Chr.12: 12942263-12942263 on Build GRCh38
rs765626493 Chr.12: 12942348-12942348 on Build GRCh38
rs780706045 Chr.12: 12942281-12942281 on Build GRCh38
rs770443584 Chr.12: 12942232-12942232 on Build GRCh38
rs184053505 Chr.12: 12942366-12942366 on Build GRCh38
rs368264783 Chr.12: 12942376-12942376 on Build GRCh38
rs769372774 Chr.12: 12942299-12942299 on Build GRCh38
rs188855286 Chr.12: 12942297-12942297 on Build GRCh38
rs772108975 Chr.12: 12942326-12942326 on Build GRCh38
rs376683696 Chr.12: 12942256-12942256 on Build GRCh38
rs894591949 Chr.12: 12942275-12942275 on Build GRCh38

View Full Product Details