Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30578846-30579115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2E2
Entrez Gene ID
2961
Gene Name
general transcription factor IIE subunit 2
Gene Aliases
FE, TF2E2, TFIIE-B, TTD6
UniGene
Hs.77100
Gene Chromosome Location
Chr.8: 30578514-30658241 on Build GRCh38
Transcripts
NM_002095.4 XM_017013363.1 XM_017013364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235219 c.867C>A p.S289S Chr.8: 30578930-30578930 on Build GRCh38
2958153 c.831G>T p.L277F Chr.8: 30578966-30578966 on Build GRCh38
3784143 c.841C>T p.L281L Chr.8: 30578956-30578956 on Build GRCh38
6472545 c.795G>C p.Q265H Chr.8: 30579002-30579002 on Build GRCh38
6676684 c.782delA p.K261fs*21 Chr.8: 30579015-30579015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759028940 Chr.8: 30578881-30578881 on Build GRCh38
rs750601837 Chr.8: 30578939-30578939 on Build GRCh38
rs755095418 Chr.8: 30579080-30579080 on Build GRCh38
rs150253885 Chr.8: 30578991-30578991 on Build GRCh38
rs781418792 Chr.8: 30579065-30579067 on Build GRCh38
rs531073678 Chr.8: 30579078-30579078 on Build GRCh38
rs940722157 Chr.8: 30578870-30578870 on Build GRCh38
rs1027576082 Chr.8: 30579008-30579008 on Build GRCh38
rs762264021 Chr.8: 30579044-30579044 on Build GRCh38
rs372950161 Chr.8: 30578873-30578873 on Build GRCh38
rs932072354 Chr.8: 30578938-30578938 on Build GRCh38
rs753638821 Chr.8: 30579084-30579084 on Build GRCh38
rs771163523 Chr.8: 30578886-30578887 on Build GRCh38
rs765890251 Chr.8: 30579059-30579059 on Build GRCh38
rs771778104 Chr.8: 30578882-30578882 on Build GRCh38
rs771547259 Chr.8: 30578993-30578993 on Build GRCh38
rs777361051 Chr.8: 30578965-30578965 on Build GRCh38
rs770909330 Chr.8: 30578980-30578980 on Build GRCh38
rs755966155 Chr.8: 30578958-30578958 on Build GRCh38
rs754376326 Chr.8: 30578904-30578904 on Build GRCh38
rs201933370 Chr.8: 30578940-30578940 on Build GRCh38
rs758525253 Chr.8: 30578943-30578943 on Build GRCh38
rs762169495 Chr.8: 30578920-30578920 on Build GRCh38
rs760180975 Chr.8: 30579009-30579009 on Build GRCh38
rs778817965 Chr.8: 30578983-30578983 on Build GRCh38
rs766529243 Chr.8: 30579075-30579075 on Build GRCh38
rs764239625 Chr.8: 30578975-30578975 on Build GRCh38
rs141953164 Chr.8: 30578952-30578952 on Build GRCh38
rs757015703 Chr.8: 30579085-30579085 on Build GRCh38
rs571887654 Chr.8: 30578994-30578994 on Build GRCh38
rs761148784 Chr.8: 30578900-30578900 on Build GRCh38
rs764328600 Chr.8: 30578902-30578902 on Build GRCh38
rs751070215 Chr.8: 30578931-30578931 on Build GRCh38
rs1042386130 Chr.8: 30579057-30579057 on Build GRCh38
rs763150502 Chr.8: 30579070-30579070 on Build GRCh38
rs776919336 Chr.8: 30579031-30579031 on Build GRCh38
rs953390125 Chr.8: 30579032-30579032 on Build GRCh38
rs750946947 Chr.8: 30579060-30579060 on Build GRCh38
rs775091073 Chr.8: 30578895-30578895 on Build GRCh38
rs769080148 Chr.8: 30579013-30579013 on Build GRCh38
rs781459162 Chr.8: 30579082-30579082 on Build GRCh38
rs371705848 Chr.8: 30578875-30578875 on Build GRCh38

View Full Product Details