Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24564121-24564394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319
Entrez Gene ID
9856
Gene Name
KIAA0319
Gene Aliases
DYLX2, DYX2, NMIG
UniGene
Hs.26441
Gene Chromosome Location
Chr.6: 24540131-24646211 on Build GRCh38
Transcripts
NM_001168374.1 NM_001168375.1 NM_001168376.1 NM_001168377.1 NM_001252328.1 NM_014809.3 XM_011515025.2 XM_011515026.2 XM_017011541.1 XM_017011542.1 XM_017011543.1 XM_017011544.1 XM_017011545.1 XM_017011546.1 XM_017011547.1 XM_017011548.1 XM_017011549.1 XM_017011550.1 XM_017011551.1 XM_017011552.1 XM_017011553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
269435 c.2398G>A p.D800N Chr.6: 24564235-24564235 on Build GRCh38
3623239 c.2301C>T p.I767I Chr.6: 24564332-24564332 on Build GRCh38
6582421 c.2403A>C p.P801P Chr.6: 24564203-24564203 on Build GRCh38
1076498 c.2396C>T p.S799L Chr.6: 24564237-24564237 on Build GRCh38
1076497 c.2404G>A p.D802N Chr.6: 24564229-24564229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377058461 Chr.6: 24564305-24564305 on Build GRCh38
rs769768664 Chr.6: 24564228-24564228 on Build GRCh38
rs374404340 Chr.6: 24564344-24564344 on Build GRCh38
rs915316764 Chr.6: 24564371-24564371 on Build GRCh38
rs375729746 Chr.6: 24564173-24564173 on Build GRCh38
rs745716352 Chr.6: 24564289-24564289 on Build GRCh38
rs754393926 Chr.6: 24564269-24564269 on Build GRCh38
rs571054596 Chr.6: 24564236-24564236 on Build GRCh38
rs375502132 Chr.6: 24564177-24564177 on Build GRCh38
rs370942417 Chr.6: 24564176-24564176 on Build GRCh38
rs766857346 Chr.6: 24564326-24564326 on Build GRCh38
rs900657478 Chr.6: 24564297-24564297 on Build GRCh38
rs9467233 Chr.6: 24564361-24564361 on Build GRCh38
rs755703736 Chr.6: 24564359-24564359 on Build GRCh38
rs750662945 Chr.6: 24564317-24564317 on Build GRCh38
rs770651150 Chr.6: 24564284-24564284 on Build GRCh38
rs775197103 Chr.6: 24564294-24564294 on Build GRCh38
rs2744550 Chr.6: 24564316-24564316 on Build GRCh38
rs570010479 Chr.6: 24564290-24564290 on Build GRCh38
rs755377473 Chr.6: 24564279-24564279 on Build GRCh38
rs998440197 Chr.6: 24564182-24564182 on Build GRCh38
rs767807604 Chr.6: 24564253-24564253 on Build GRCh38
rs780765884 Chr.6: 24564266-24564266 on Build GRCh38
rs775390095 Chr.6: 24564232-24564232 on Build GRCh38
rs1026181590 Chr.6: 24564324-24564324 on Build GRCh38
rs749098866 Chr.6: 24564360-24564360 on Build GRCh38
rs746459020 Chr.6: 24564283-24564283 on Build GRCh38
rs747594850 Chr.6: 24564216-24564217 on Build GRCh38
rs761190759 Chr.6: 24564318-24564318 on Build GRCh38
rs375534519 Chr.6: 24564188-24564188 on Build GRCh38
rs751126414 Chr.6: 24564348-24564348 on Build GRCh38
rs2760164 Chr.6: 24564345-24564345 on Build GRCh38
rs2817191 Chr.6: 24564312-24564312 on Build GRCh38
rs751533886 Chr.6: 24564240-24564240 on Build GRCh38
rs201132279 Chr.6: 24564245-24564245 on Build GRCh38
rs1032784188 Chr.6: 24564261-24564261 on Build GRCh38
rs200477022 Chr.6: 24564372-24564372 on Build GRCh38
rs146274466 Chr.6: 24564362-24564362 on Build GRCh38
rs778578541 Chr.6: 24564164-24564165 on Build GRCh38
rs199999798 Chr.6: 24564304-24564304 on Build GRCh38
rs759292346 Chr.6: 24564225-24564225 on Build GRCh38
rs754096986 Chr.6: 24564332-24564332 on Build GRCh38
rs764238150 Chr.6: 24564237-24564237 on Build GRCh38
rs776436431 Chr.6: 24564224-24564224 on Build GRCh38
rs748319045 Chr.6: 24564373-24564373 on Build GRCh38
rs756526138 Chr.6: 24564262-24564262 on Build GRCh38
rs771881910 Chr.6: 24564218-24564225 on Build GRCh38
rs2760163 Chr.6: 24564351-24564351 on Build GRCh38
rs780797707 Chr.6: 24564288-24564288 on Build GRCh38
rs756071583 Chr.6: 24564355-24564355 on Build GRCh38
rs768679258 Chr.6: 24564366-24564366 on Build GRCh38
rs376652269 Chr.6: 24564189-24564189 on Build GRCh38
rs770436022 Chr.6: 24564218-24564218 on Build GRCh38
rs201872640 Chr.6: 24564169-24564169 on Build GRCh38
rs149012243 Chr.6: 24564296-24564296 on Build GRCh38
rs777359139 Chr.6: 24564280-24564280 on Build GRCh38
rs550491204 Chr.6: 24564251-24564251 on Build GRCh38
rs371137479 Chr.6: 24564259-24564259 on Build GRCh38
rs374727009 Chr.6: 24564354-24564354 on Build GRCh38

View Full Product Details