Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166198948-166199191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507589 c.5546G>T p.R1849L Chr.2: 166199096-166199096 on Build GRCh38
5394777 c.5467C>T p.Q1823* Chr.2: 166199175-166199175 on Build GRCh38
3406998 c.5482G>T p.D1828Y Chr.2: 166199160-166199160 on Build GRCh38
3569139 c.5625G>A p.R1875R Chr.2: 166198981-166198981 on Build GRCh38
4763399 c.5468A>C p.Q1823P Chr.2: 166199174-166199174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762575742 Chr.2: 166199151-166199151 on Build GRCh38
rs751325279 Chr.2: 166199152-166199152 on Build GRCh38
rs749373098 Chr.2: 166199077-166199077 on Build GRCh38
rs769295581 Chr.2: 166199135-166199135 on Build GRCh38
rs529045323 Chr.2: 166199009-166199009 on Build GRCh38
rs879254298 Chr.2: 166199024-166199024 on Build GRCh38
rs199705100 Chr.2: 166199096-166199096 on Build GRCh38
rs200803311 Chr.2: 166199149-166199149 on Build GRCh38
rs761344025 Chr.2: 166199049-166199049 on Build GRCh38
rs751361718 Chr.2: 166199069-166199069 on Build GRCh38
rs761434362 Chr.2: 166199141-166199141 on Build GRCh38
rs367719041 Chr.2: 166198975-166198975 on Build GRCh38
rs200478305 Chr.2: 166199008-166199008 on Build GRCh38
rs747340707 Chr.2: 166199131-166199131 on Build GRCh38
rs876661011 Chr.2: 166199078-166199078 on Build GRCh38
rs1026656279 Chr.2: 166199098-166199098 on Build GRCh38
rs895014544 Chr.2: 166198989-166198989 on Build GRCh38
rs560168893 Chr.2: 166198983-166198983 on Build GRCh38
rs867949537 Chr.2: 166199038-166199038 on Build GRCh38
rs766124857 Chr.2: 166199060-166199060 on Build GRCh38
rs761153502 Chr.2: 166199016-166199016 on Build GRCh38
rs754924014 Chr.2: 166199072-166199072 on Build GRCh38
rs201035087 Chr.2: 166198972-166198972 on Build GRCh38
rs781049315 Chr.2: 166199073-166199073 on Build GRCh38
rs759259640 Chr.2: 166199165-166199165 on Build GRCh38
rs772335195 Chr.2: 166199118-166199118 on Build GRCh38
rs199572382 Chr.2: 166199053-166199053 on Build GRCh38
rs201483184 Chr.2: 166199084-166199084 on Build GRCh38
rs764685430 Chr.2: 166199025-166199025 on Build GRCh38
rs201561928 Chr.2: 166199051-166199051 on Build GRCh38
rs200608140 Chr.2: 166199136-166199136 on Build GRCh38
rs776299330 Chr.2: 166199037-166199037 on Build GRCh38
rs371379751 Chr.2: 166199112-166199112 on Build GRCh38
rs201972510 Chr.2: 166199166-166199166 on Build GRCh38
rs201638678 Chr.2: 166199142-166199142 on Build GRCh38
rs202239457 Chr.2: 166198973-166198973 on Build GRCh38
rs776271263 Chr.2: 166199024-166199024 on Build GRCh38
rs746119070 Chr.2: 166199116-166199116 on Build GRCh38
rs200196731 Chr.2: 166199148-166199148 on Build GRCh38
rs779128646 Chr.2: 166199114-166199114 on Build GRCh38
rs775777109 Chr.2: 166199127-166199127 on Build GRCh38
rs774488202 Chr.2: 166198982-166198982 on Build GRCh38
rs772373532 Chr.2: 166198986-166198986 on Build GRCh38
rs757285775 Chr.2: 166199087-166199087 on Build GRCh38
rs200494490 Chr.2: 166199061-166199061 on Build GRCh38
rs876661248 Chr.2: 166199076-166199076 on Build GRCh38

View Full Product Details