The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000972.2 | NP_000963.1 | 6 - 7 | 657 | 64 |
GenBank mRNA | AB056665.1 | - | 3 - 4 | 256 | 64 |
AK222534.1 | - | 6 - 7 | 654 | 64 | |
AK291123.1 | - | 6 - 7 | 649 | 64 | |
AK311743.1 | - | 6 - 7 | 653 | 64 | |
BC005128.1 | - | 1 - 1 | 640 | 64 | |
BC021052.1 | - | 1 - 1 | 631 | 64 | |
BC021979.2 | - | 1 - 1 | 629 | 64 | |
BC023594.2 | - | 6 - 7 | 688 | 64 | |
BC023624.2 | - | 6 - 7 | 693 | 64 | |
BC032533.1 | - | 1 - 1 | 676 | 64 | |
BC071900.1 | - | 1 - 1 | 629 | 64 | |
BC071901.1 | - | 5 - 6 | 637 | 64 | |
BC073802.1 | - | 1 - 1 | 628 | 64 | |
BC105290.1 | - | 1 - 1 | 654 | 64 | |
M36072.1 | - | 6 - 7 | 658 | 64 | |
X06705.1 | - | 6 - 7 | 657 | 64 |