Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38120774-38121025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_011530426.2 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1 XM_017028986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3554346 c.1649G>A p.R550Q Chr.22: 38120852-38120852 on Build GRCh38
6747779 c.1724C>T p.T575M Chr.22: 38120777-38120777 on Build GRCh38
5057812 c.1615G>A p.G539S Chr.22: 38120886-38120886 on Build GRCh38
3554345 c.1677C>T p.P559P Chr.22: 38120824-38120824 on Build GRCh38
5003129 c.1632G>T p.M544I Chr.22: 38120869-38120869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763937995 Chr.22: 38120873-38120873 on Build GRCh38
rs952349435 Chr.22: 38120863-38120863 on Build GRCh38
rs200055035 Chr.22: 38120945-38120945 on Build GRCh38
rs769047790 Chr.22: 38120831-38120831 on Build GRCh38
rs748730147 Chr.22: 38120901-38120901 on Build GRCh38
rs149015861 Chr.22: 38120818-38120818 on Build GRCh38
rs535486098 Chr.22: 38120888-38120888 on Build GRCh38
rs928761158 Chr.22: 38120984-38120984 on Build GRCh38
rs1004616610 Chr.22: 38120853-38120853 on Build GRCh38
rs761769964 Chr.22: 38120935-38120936 on Build GRCh38
rs887630274 Chr.22: 38120824-38120824 on Build GRCh38
rs946244566 Chr.22: 38120963-38120963 on Build GRCh38
rs757250238 Chr.22: 38120880-38120880 on Build GRCh38
rs752452448 Chr.22: 38120866-38120866 on Build GRCh38
rs537635198 Chr.22: 38120965-38120965 on Build GRCh38
rs777259654 Chr.22: 38120835-38120835 on Build GRCh38
rs764002301 Chr.22: 38120932-38120932 on Build GRCh38
rs146415291 Chr.22: 38120812-38120812 on Build GRCh38
rs587784336 Chr.22: 38120827-38120827 on Build GRCh38
rs527807604 Chr.22: 38120881-38120881 on Build GRCh38
rs540519438 Chr.22: 38121002-38121002 on Build GRCh38
rs565901019 Chr.22: 38120922-38120922 on Build GRCh38
rs576759811 Chr.22: 38120964-38120964 on Build GRCh38
rs752382392 Chr.22: 38120992-38120992 on Build GRCh38
rs1027463904 Chr.22: 38120806-38120806 on Build GRCh38
rs770478201 Chr.22: 38120836-38120836 on Build GRCh38
rs751508677 Chr.22: 38120816-38120816 on Build GRCh38
rs776015045 Chr.22: 38120827-38120827 on Build GRCh38
rs767273844 Chr.22: 38120991-38120991 on Build GRCh38
rs370691849 Chr.22: 38120889-38120889 on Build GRCh38
rs121908681 Chr.22: 38120867-38120867 on Build GRCh38
rs143826762 Chr.22: 38120886-38120886 on Build GRCh38
rs772522384 Chr.22: 38120822-38120822 on Build GRCh38
rs895050694 Chr.22: 38120989-38120989 on Build GRCh38
rs587784337 Chr.22: 38120802-38120802 on Build GRCh38
rs759132756 Chr.22: 38120915-38120915 on Build GRCh38
rs747491292 Chr.22: 38120830-38120830 on Build GRCh38
rs1041768594 Chr.22: 38120933-38120933 on Build GRCh38
rs755719764 Chr.22: 38121006-38121006 on Build GRCh38
rs750534315 Chr.22: 38120953-38120953 on Build GRCh38
rs760379057 Chr.22: 38120928-38120928 on Build GRCh38
rs770388313 Chr.22: 38120909-38120909 on Build GRCh38
rs374315286 Chr.22: 38120851-38120851 on Build GRCh38
rs376051062 Chr.22: 38120852-38120852 on Build GRCh38
rs758616263 Chr.22: 38120955-38120955 on Build GRCh38
rs771719117 Chr.22: 38120918-38120918 on Build GRCh38
rs773784597 Chr.22: 38120983-38120983 on Build GRCh38
rs769122831 Chr.22: 38120891-38120891 on Build GRCh38
rs867819529 Chr.22: 38120846-38120846 on Build GRCh38
rs773961836 Chr.22: 38120849-38120849 on Build GRCh38
rs918251529 Chr.22: 38120857-38120857 on Build GRCh38
rs886057502 Chr.22: 38120853-38120853 on Build GRCh38
rs773875677 Chr.22: 38120913-38120913 on Build GRCh38
rs183978140 Chr.22: 38120893-38120893 on Build GRCh38
rs373930150 Chr.22: 38120887-38120887 on Build GRCh38
rs753874848 Chr.22: 38120874-38120874 on Build GRCh38
rs780125490 Chr.22: 38120958-38120958 on Build GRCh38
rs549902731 Chr.22: 38120805-38120805 on Build GRCh38
rs558124373 Chr.22: 38120946-38120946 on Build GRCh38

View Full Product Details