Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIH1D3
Entrez Gene ID
139212
Gene Name
PIH1 domain containing 3
Gene Aliases
CXorf41, NYSAR97
Chromosome Location
Chr.X: 107206629 - 107244243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173494.1 NP_775765.1 1 - 2 51 136
GenBank mRNA BC033510.1 - 1 - 2 51 136

View Full Product Details