Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19orf70
Entrez Gene ID
125988
Gene Name
chromosome 19 open reading frame 70
Gene Aliases
MIC13, P117, QIL1
Chromosome Location
Chr.19: 5678414 - 5680896 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308240.1 NP_001295169.1 4 - 5 864 80
NM_205767.2 NP_991330.1 3 - 4 668 80
XM_017026248.1 XP_016881737.1 3 - 4 609 80
GenBank mRNA AA292888.1 - 3 - 4 435 80
AI300708.1 - 1 - 2 80
AJ707830.1 - 3 - 4 414 80
BC009557.1 - 3 - 4 302 80
BC042386.1 - 3 - 4 277 80
BC052640.1 - 3 - 4 279 80
BI827162.1 - 3 - 4 667 80

View Full Product Details