Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96131670-96131943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A13
Entrez Gene ID
10165
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
UniGene
Hs.489190
Gene Chromosome Location
Chr.7: 96120220-96322147 on Build GRCh38
Transcripts
NM_001160210.1 NM_014251.2 NR_027662.1 XM_006715831.3 XM_017011663.1 XM_017011664.1 XM_017011665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5497385 c.1565G>C p.S522T Chr.7: 96131769-96131769 on Build GRCh38
5680947 c.1466G>T p.C489F Chr.7: 96131868-96131868 on Build GRCh38
4898544 c.1529C>T p.S510F Chr.7: 96131805-96131805 on Build GRCh38
5985010 c.1474C>T p.R492W Chr.7: 96131860-96131860 on Build GRCh38
3642602 c.1562G>A p.G521E Chr.7: 96131772-96131772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375405382 Chr.7: 96131748-96131748 on Build GRCh38
rs745628778 Chr.7: 96131831-96131831 on Build GRCh38
rs774001538 Chr.7: 96131739-96131739 on Build GRCh38
rs752235032 Chr.7: 96131799-96131799 on Build GRCh38
rs761992118 Chr.7: 96131851-96131851 on Build GRCh38
rs191239117 Chr.7: 96131885-96131885 on Build GRCh38
rs753258126 Chr.7: 96131917-96131917 on Build GRCh38
rs775532487 Chr.7: 96131843-96131843 on Build GRCh38
rs755432934 Chr.7: 96131697-96131697 on Build GRCh38
rs369962634 Chr.7: 96131796-96131796 on Build GRCh38
rs756599320 Chr.7: 96131906-96131906 on Build GRCh38
rs771908277 Chr.7: 96131768-96131768 on Build GRCh38
rs961627223 Chr.7: 96131754-96131754 on Build GRCh38
rs750844927 Chr.7: 96131693-96131693 on Build GRCh38
rs186863471 Chr.7: 96131830-96131830 on Build GRCh38
rs1034142358 Chr.7: 96131708-96131708 on Build GRCh38
rs532545471 Chr.7: 96131706-96131706 on Build GRCh38
rs750610043 Chr.7: 96131786-96131786 on Build GRCh38
rs765591535 Chr.7: 96131784-96131784 on Build GRCh38
rs780718018 Chr.7: 96131703-96131703 on Build GRCh38
rs538166755 Chr.7: 96131898-96131898 on Build GRCh38
rs886605906 Chr.7: 96131729-96131729 on Build GRCh38
rs770700025 Chr.7: 96131730-96131730 on Build GRCh38
rs751973213 Chr.7: 96131884-96131884 on Build GRCh38
rs200004471 Chr.7: 96131904-96131904 on Build GRCh38
rs777329706 Chr.7: 96131914-96131914 on Build GRCh38
rs747495339 Chr.7: 96131696-96131696 on Build GRCh38
rs781625058 Chr.7: 96131798-96131798 on Build GRCh38
rs1013572793 Chr.7: 96131732-96131732 on Build GRCh38
rs769939259 Chr.7: 96131859-96131859 on Build GRCh38
rs1002312963 Chr.7: 96131700-96131700 on Build GRCh38
rs139455686 Chr.7: 96131766-96131766 on Build GRCh38
rs144494809 Chr.7: 96131828-96131828 on Build GRCh38
rs756182703 Chr.7: 96131845-96131848 on Build GRCh38
rs763272772 Chr.7: 96131860-96131860 on Build GRCh38
rs181566129 Chr.7: 96131694-96131694 on Build GRCh38
rs777414201 Chr.7: 96131823-96131823 on Build GRCh38
rs963663111 Chr.7: 96131809-96131809 on Build GRCh38
rs375858610 Chr.7: 96131712-96131712 on Build GRCh38
rs766485284 Chr.7: 96131869-96131869 on Build GRCh38
rs139149160 Chr.7: 96131829-96131829 on Build GRCh38
rs11558671 Chr.7: 96131815-96131815 on Build GRCh38
rs749039874 Chr.7: 96131724-96131724 on Build GRCh38
rs772157920 Chr.7: 96131837-96131837 on Build GRCh38
rs944437173 Chr.7: 96131903-96131903 on Build GRCh38
rs763244253 Chr.7: 96131787-96131787 on Build GRCh38
rs751836823 Chr.7: 96131795-96131795 on Build GRCh38
rs772811349 Chr.7: 96131721-96131721 on Build GRCh38
rs772104865 Chr.7: 96131749-96131749 on Build GRCh38
rs1005040991 Chr.7: 96131746-96131746 on Build GRCh38
rs766827198 Chr.7: 96131790-96131790 on Build GRCh38
rs373291117 Chr.7: 96131695-96131695 on Build GRCh38
rs765534313 Chr.7: 96131692-96131692 on Build GRCh38

View Full Product Details