Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF69
Entrez Gene ID
7620
Gene Name
zinc finger protein 69
Gene Aliases
Cos5, hZNF3
Chromosome Location
Chr.19: 11887773 - 11980222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021915.3 NP_068734.1 1 - 2 202 124
GenBank mRNA BC051309.1 - 1 - 2 131 124

View Full Product Details