The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303136.1 | NP_001290065.1 | 19 - 20 | 2954 | 110 |
NM_152753.3 | NP_689966.2 | 19 - 20 | 2957 | 110 | |
XM_005248943.1 | XP_005249000.1 | 20 - 21 | 3020 | 110 | |
XM_005248947.1 | XP_005249004.1 | 19 - 20 | 2858 | 110 | |
XM_011514404.2 | XP_011512706.1 | 20 - 21 | 2720 | 110 | |
GenBank mRNA | AF452494.1 | - | 19 - 20 | 2606 | 110 |
AK092010.1 | - | 11 - 12 | 1610 | 110 | |
AK092062.1 | - | 19 - 20 | 2516 | 110 | |
AK291218.1 | - | 20 - 21 | 2678 | 110 | |
AY639608.1 | - | 19 - 20 | 2600 | 110 | |
BC052263.1 | - | 19 - 20 | 2954 | 110 | |
CR936607.1 | - | 16 - 17 | 2178 | 110 |