The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033159.3 | NP_149349.2 | 1 - 2 | 135 | 94 |
NM_153282.2 | NP_695014.1 | 1 - 2 | 135 | 94 | |
GenBank mRNA | AF118821.1 | - | 1 - 2 | 100 | 94 |
AF173154.1 | - | 1 - 2 | 106 | 94 | |
AF502904.1 | - | 1 - 2 | 107 | 94 | |
AF502906.1 | - | 1 - 2 | 107 | 94 | |
AK290816.1 | - | 1 - 2 | 127 | 94 | |
BC025774.1 | - | 1 - 2 | 83 | 94 | |
BC035695.1 | - | 1 - 2 | 89 | 94 | |
DA566755.1 | - | 1 - 2 | 137 | 94 |