Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144512329-144512566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569133 c.3014G>A p.R1005Q Chr.8: 144512433-144512433 on Build GRCh38
2871877 c.2987T>C p.M996T Chr.8: 144512460-144512460 on Build GRCh38
6764678 c.3055G>T p.G1019C Chr.8: 144512392-144512392 on Build GRCh38
6963891 c.3011G>A p.R1004Q Chr.8: 144512436-144512436 on Build GRCh38
6924491 c.3031_3032insTGC p.L1010_Q1011insL Chr.8: 144512415-144512416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775795557 Chr.8: 144512471-144512471 on Build GRCh38
rs1008463415 Chr.8: 144512376-144512376 on Build GRCh38
rs772780752 Chr.8: 144512540-144512540 on Build GRCh38
rs372511135 Chr.8: 144512385-144512385 on Build GRCh38
rs746107365 Chr.8: 144512514-144512514 on Build GRCh38
rs779017449 Chr.8: 144512386-144512386 on Build GRCh38
rs758067843 Chr.8: 144512364-144512364 on Build GRCh38
rs368585080 Chr.8: 144512391-144512391 on Build GRCh38
rs369172759 Chr.8: 144512449-144512449 on Build GRCh38
rs781661324 Chr.8: 144512461-144512461 on Build GRCh38
rs903820228 Chr.8: 144512349-144512349 on Build GRCh38
rs377031190 Chr.8: 144512387-144512387 on Build GRCh38
rs771581851 Chr.8: 144512537-144512537 on Build GRCh38
rs373784742 Chr.8: 144512495-144512495 on Build GRCh38
rs587778645 Chr.8: 144512512-144512512 on Build GRCh38
rs200018416 Chr.8: 144512480-144512480 on Build GRCh38
rs751727270 Chr.8: 144512498-144512498 on Build GRCh38
rs760402848 Chr.8: 144512541-144512541 on Build GRCh38
rs767220757 Chr.8: 144512481-144512481 on Build GRCh38
rs776993029 Chr.8: 144512505-144512506 on Build GRCh38
rs760096494 Chr.8: 144512407-144512407 on Build GRCh38
rs980962861 Chr.8: 144512462-144512462 on Build GRCh38
rs758339361 Chr.8: 144512447-144512447 on Build GRCh38
rs770014424 Chr.8: 144512515-144512515 on Build GRCh38
rs757541591 Chr.8: 144512506-144512506 on Build GRCh38
rs151223648 Chr.8: 144512479-144512479 on Build GRCh38
rs767281689 Chr.8: 144512435-144512435 on Build GRCh38
rs368982201 Chr.8: 144512434-144512434 on Build GRCh38
rs762312510 Chr.8: 144512425-144512425 on Build GRCh38
rs898859819 Chr.8: 144512409-144512409 on Build GRCh38
rs750261857 Chr.8: 144512436-144512436 on Build GRCh38
rs780492921 Chr.8: 144512518-144512518 on Build GRCh38
rs778030723 Chr.8: 144512448-144512448 on Build GRCh38
rs902652277 Chr.8: 144512450-144512450 on Build GRCh38
rs771326102 Chr.8: 144512460-144512460 on Build GRCh38
rs35070885 Chr.8: 144512521-144512521 on Build GRCh38
rs764754383 Chr.8: 144512358-144512358 on Build GRCh38
rs36023964 Chr.8: 144512437-144512437 on Build GRCh38
rs750400159 Chr.8: 144512405-144512405 on Build GRCh38
rs771381171 Chr.8: 144512404-144512404 on Build GRCh38
rs760150078 Chr.8: 144512357-144512357 on Build GRCh38
rs999814906 Chr.8: 144512456-144512456 on Build GRCh38
rs200629599 Chr.8: 144512494-144512494 on Build GRCh38
rs750202919 Chr.8: 144512482-144512482 on Build GRCh38
rs745759208 Chr.8: 144512379-144512379 on Build GRCh38
rs373202723 Chr.8: 144512464-144512464 on Build GRCh38
rs4251691 Chr.8: 144512433-144512433 on Build GRCh38
rs770209478 Chr.8: 144512465-144512465 on Build GRCh38
rs761367235 Chr.8: 144512534-144512534 on Build GRCh38
rs756983251 Chr.8: 144512372-144512372 on Build GRCh38
rs763712794 Chr.8: 144512367-144512367 on Build GRCh38
rs182454890 Chr.8: 144512368-144512368 on Build GRCh38
rs766197579 Chr.8: 144512489-144512489 on Build GRCh38
rs372660887 Chr.8: 144512420-144512420 on Build GRCh38
rs772320981 Chr.8: 144512388-144512388 on Build GRCh38
rs773849437 Chr.8: 144512525-144512525 on Build GRCh38
rs34919378 Chr.8: 144512438-144512438 on Build GRCh38
rs769194048 Chr.8: 144512523-144512523 on Build GRCh38
rs763547531 Chr.8: 144512474-144512474 on Build GRCh38
rs747239208 Chr.8: 144512398-144512398 on Build GRCh38
rs918047641 Chr.8: 144512445-144512445 on Build GRCh38
rs760325940 Chr.8: 144512484-144512484 on Build GRCh38
rs763665817 Chr.8: 144512431-144512431 on Build GRCh38
rs777976493 Chr.8: 144512496-144512496 on Build GRCh38
rs948234493 Chr.8: 144512458-144512458 on Build GRCh38
rs765732606 Chr.8: 144512415-144512415 on Build GRCh38
rs781000093 Chr.8: 144512375-144512375 on Build GRCh38
rs752195344 Chr.8: 144512359-144512359 on Build GRCh38

View Full Product Details