The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202543.1 | NP_001189472.1 | 16 - 17 | 2121 | 99 |
NM_181552.3 | NP_853530.2 | 16 - 17 | 1999 | 99 | |
XM_005250150.2 | XP_005250207.1 | 16 - 17 | 2520 | 99 | |
XM_005250151.2 | XP_005250208.1 | 16 - 17 | 2517 | 99 | |
XM_011515823.2 | XP_011514125.1 | 16 - 17 | 2212 | 99 | |
XM_011515825.1 | XP_011514127.1 | 16 - 17 | 1780 | 99 | |
XM_017011759.1 | XP_016867248.1 | 16 - 17 | 2080 | 99 | |
GenBank mRNA | BC066592.1 | - | 16 - 17 | 2014 | 99 |
M74099.1 | - | 16 - 17 | 2004 | 99 |