Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CUX1
- Entrez Gene ID
-
1523
- Gene Name
- cut like homeobox 1
- Gene Aliases
-
CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75
- Chromosome Location
-
Chr.7: 101815904 - 102283958 on Build GRCh38
View Full Product Details