The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202543.1 | NP_001189472.1 | 11 - 12 | 1175 | 63 |
NM_001202544.2 | NP_001189473.1 | 10 - 11 | 1127 | 63 | |
NM_001202545.2 | NP_001189474.1 | 10 - 11 | 1037 | 63 | |
NM_001202546.2 | NP_001189475.1 | 10 - 11 | 954 | 63 | |
NM_001913.4 | NP_001904.2 | 11 - 12 | 1175 | 63 | |
NM_181500.3 | NP_852477.1 | 11 - 12 | 1169 | 63 | |
NM_181552.3 | NP_853530.2 | 11 - 12 | 1053 | 63 | |
XM_005250150.2 | XP_005250207.1 | 11 - 12 | 1574 | 63 | |
XM_005250151.2 | XP_005250208.1 | 11 - 12 | 1571 | 63 | |
XM_005250154.3 | XP_005250211.1 | 11 - 12 | 1431 | 63 | |
XM_006715854.2 | XP_006715917.1 | 11 - 12 | 1574 | 63 | |
XM_006715855.1 | XP_006715918.1 | 11 - 12 | 1425 | 63 | |
XM_011515823.2 | XP_011514125.1 | 11 - 12 | 1572 | 63 | |
XM_011515825.1 | XP_011514127.1 | 11 - 12 | 1140 | 63 | |
XM_017011759.1 | XP_016867248.1 | 11 - 12 | 1134 | 63 | |
XM_017011760.1 | XP_016867249.1 | 11 - 12 | 1134 | 63 | |
GenBank mRNA | AB075522.1 | - | 8 - 9 | 827 | 63 |
AK122726.1 | - | 10 - 11 | 920 | 63 | |
AK125097.1 | - | 10 - 11 | 1022 | 63 | |
AK222832.1 | - | 11 - 12 | 1071 | 63 | |
AK303151.1 | - | 10 - 11 | 958 | 63 | |
BC025422.2 | - | 11 - 12 | 1050 | 63 | |
BC066592.1 | - | 11 - 12 | 1068 | 63 | |
L12579.1 | - | 11 - 12 | 1067 | 63 | |
M74099.1 | - | 11 - 12 | 1058 | 63 |