Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14848510-14848735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
UniGene
Hs.534169
Gene Chromosome Location
Chr.10: 14838160-14871741 on Build GRCh38
Transcripts
NM_016299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465481 c.262A>C p.K88Q Chr.10: 14848649-14848649 on Build GRCh38
23329 c.254C>T p.A85V Chr.10: 14848641-14848641 on Build GRCh38
5999094 c.229A>G p.K77E Chr.10: 14848650-14848650 on Build GRCh38
4012977 c.253G>A p.A85T Chr.10: 14848640-14848640 on Build GRCh38
916598 c.252C>T p.I84I Chr.10: 14848639-14848639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550196167 Chr.10: 14848642-14848642 on Build GRCh38
rs540257564 Chr.10: 14848689-14848689 on Build GRCh38
rs780019332 Chr.10: 14848564-14848564 on Build GRCh38
rs771702547 Chr.10: 14848585-14848585 on Build GRCh38
rs754882053 Chr.10: 14848682-14848682 on Build GRCh38
rs781620850 Chr.10: 14848559-14848559 on Build GRCh38
rs374837716 Chr.10: 14848641-14848641 on Build GRCh38
rs893717664 Chr.10: 14848556-14848556 on Build GRCh38
rs764472017 Chr.10: 14848583-14848583 on Build GRCh38
rs114573476 Chr.10: 14848568-14848568 on Build GRCh38
rs753738915 Chr.10: 14848643-14848643 on Build GRCh38
rs150852688 Chr.10: 14848669-14848670 on Build GRCh38
rs201822829 Chr.10: 14848692-14848692 on Build GRCh38
rs577529675 Chr.10: 14848684-14848684 on Build GRCh38
rs777386710 Chr.10: 14848691-14848691 on Build GRCh38
rs199859571 Chr.10: 14848640-14848640 on Build GRCh38
rs375004850 Chr.10: 14848639-14848639 on Build GRCh38
rs186835892 Chr.10: 14848704-14848704 on Build GRCh38
rs751957518 Chr.10: 14848625-14848625 on Build GRCh38
rs747935446 Chr.10: 14848686-14848686 on Build GRCh38
rs768454712 Chr.10: 14848576-14848576 on Build GRCh38
rs181803932 Chr.10: 14848601-14848601 on Build GRCh38
rs777799732 Chr.10: 14848588-14848589 on Build GRCh38
rs764455012 Chr.10: 14848622-14848622 on Build GRCh38
rs747743019 Chr.10: 14848584-14848584 on Build GRCh38
rs772918903 Chr.10: 14848593-14848593 on Build GRCh38
rs776017817 Chr.10: 14848606-14848606 on Build GRCh38
rs1015031510 Chr.10: 14848628-14848628 on Build GRCh38
rs756187884 Chr.10: 14848561-14848561 on Build GRCh38
rs897268037 Chr.10: 14848652-14848652 on Build GRCh38
rs573121763 Chr.10: 14848560-14848560 on Build GRCh38
rs763402880 Chr.10: 14848619-14848619 on Build GRCh38
rs188358184 Chr.10: 14848581-14848581 on Build GRCh38
rs770871363 Chr.10: 14848686-14848689 on Build GRCh38
rs184018354 Chr.10: 14848558-14848558 on Build GRCh38
rs760151001 Chr.10: 14848600-14848600 on Build GRCh38

View Full Product Details