The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005233.5 | NP_005224.2 | 3 - 4 | 1034 | 73 |
NM_182644.2 | NP_872585.1 | 3 - 4 | 1034 | 73 | |
XM_005264715.2 | XP_005264772.1 | 3 - 4 | 959 | 73 | |
XM_005264716.2 | XP_005264773.1 | 3 - 4 | 1029 | 73 | |
GenBank mRNA | AF213459.1 | - | 3 - 4 | 1034 | 73 |
AF213460.1 | - | 3 - 4 | 896 | 73 | |
AK291411.1 | - | 3 - 4 | 867 | 73 | |
BC063282.1 | - | 3 - 4 | 938 | 73 | |
M83941.1 | - | 3 - 4 | 909 | 73 |