The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006936.2 | NP_008867.2 | 2 - 3 | 313 | 149 |
GenBank mRNA | AK222591.1 | - | 2 - 3 | 244 | 149 |
AK312350.1 | - | 2 - 3 | 244 | 149 | |
BC000036.2 | - | 2 - 3 | 238 | 149 | |
BC008420.1 | - | 2 - 3 | 313 | 149 | |
BT007008.1 | - | 2 - 3 | 152 | 149 | |
CR542173.1 | - | 2 - 3 | 152 | 149 | |
CR542188.1 | - | 2 - 3 | 152 | 149 | |
X99584.1 | - | 2 - 3 | 246 | 149 |