Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6414019-6414278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHSRP
Entrez Gene ID
8570
Gene Name
KH-type splicing regulatory protein
Gene Aliases
FBP2, FUBP2, KSRP
UniGene
Hs.727344
Gene Chromosome Location
Chr.19: 6413104-6424811 on Build GRCh38
Transcripts
NM_003685.2 XM_005259668.4 XM_011528395.2 XM_017027408.1 XM_017027409.1 XM_017027410.1

SNP Detail

SNP ID Chromosome Location
rs532530916 Chr.19: 6414156-6414156 on Build GRCh38
rs369293348 Chr.19: 6414075-6414075 on Build GRCh38
rs757003676 Chr.19: 6414186-6414186 on Build GRCh38
rs750674142 Chr.19: 6414094-6414094 on Build GRCh38
rs748645919 Chr.19: 6414255-6414255 on Build GRCh38
rs767612961 Chr.19: 6414218-6414218 on Build GRCh38
rs746454398 Chr.19: 6414198-6414198 on Build GRCh38
rs549829161 Chr.19: 6414184-6414184 on Build GRCh38
rs780892331 Chr.19: 6414252-6414252 on Build GRCh38
rs752352009 Chr.19: 6414248-6414248 on Build GRCh38
rs767992437 Chr.19: 6414196-6414196 on Build GRCh38
rs766000184 Chr.19: 6414245-6414245 on Build GRCh38
rs185800143 Chr.19: 6414244-6414244 on Build GRCh38
rs1046956461 Chr.19: 6414042-6414042 on Build GRCh38
rs766514395 Chr.19: 6414109-6414109 on Build GRCh38
rs769600035 Chr.19: 6414065-6414065 on Build GRCh38
rs35196078 Chr.19: 6414219-6414220 on Build GRCh38
rs756096840 Chr.19: 6414124-6414124 on Build GRCh38
rs771631974 Chr.19: 6414065-6414065 on Build GRCh38
rs968125243 Chr.19: 6414162-6414162 on Build GRCh38
rs533282866 Chr.19: 6414170-6414170 on Build GRCh38
rs762981895 Chr.19: 6414065-6414066 on Build GRCh38
rs781746819 Chr.19: 6414205-6414205 on Build GRCh38
rs774500044 Chr.19: 6414089-6414091 on Build GRCh38
rs748472349 Chr.19: 6414181-6414181 on Build GRCh38
rs529449095 Chr.19: 6414214-6414214 on Build GRCh38
rs986016464 Chr.19: 6414240-6414240 on Build GRCh38
rs754500164 Chr.19: 6414250-6414250 on Build GRCh38
rs762008747 Chr.19: 6414178-6414179 on Build GRCh38
rs778379074 Chr.19: 6414143-6414143 on Build GRCh38
rs200942635 Chr.19: 6414155-6414155 on Build GRCh38
rs777283442 Chr.19: 6414161-6414161 on Build GRCh38
rs569936769 Chr.19: 6414210-6414210 on Build GRCh38
rs766350050 Chr.19: 6414174-6414174 on Build GRCh38
rs777535739 Chr.19: 6414125-6414125 on Build GRCh38
rs770307807 Chr.19: 6414166-6414166 on Build GRCh38
rs762122897 Chr.19: 6414091-6414091 on Build GRCh38
rs750720082 Chr.19: 6414210-6414211 on Build GRCh38
rs560194408 Chr.19: 6414139-6414139 on Build GRCh38
rs748698842 Chr.19: 6414209-6414209 on Build GRCh38
rs369861500 Chr.19: 6414164-6414164 on Build GRCh38
rs779424955 Chr.19: 6414196-6414196 on Build GRCh38
rs745563975 Chr.19: 6414152-6414152 on Build GRCh38
rs753741621 Chr.19: 6414219-6414219 on Build GRCh38
rs761786190 Chr.19: 6414229-6414229 on Build GRCh38
rs541460193 Chr.19: 6414115-6414115 on Build GRCh38
rs775818254 Chr.19: 6414069-6414069 on Build GRCh38
rs777193847 Chr.19: 6414077-6414077 on Build GRCh38
rs759560197 Chr.19: 6414217-6414217 on Build GRCh38
rs768870967 Chr.19: 6414065-6414067 on Build GRCh38
rs773679047 Chr.19: 6414212-6414212 on Build GRCh38
rs775569966 Chr.19: 6414064-6414065 on Build GRCh38
rs750020750 Chr.19: 6414187-6414187 on Build GRCh38
rs769353158 Chr.19: 6414159-6414159 on Build GRCh38
rs761291982 Chr.19: 6414211-6414212 on Build GRCh38
rs376723443 Chr.19: 6414093-6414093 on Build GRCh38
rs960736764 Chr.19: 6414154-6414154 on Build GRCh38
rs924126818 Chr.19: 6414179-6414179 on Build GRCh38
rs909299294 Chr.19: 6414196-6414216 on Build GRCh38
rs529055591 Chr.19: 6414185-6414185 on Build GRCh38
rs768296568 Chr.19: 6414202-6414202 on Build GRCh38
rs887002938 Chr.19: 6414073-6414073 on Build GRCh38
rs773269595 Chr.19: 6414167-6414167 on Build GRCh38
rs373531467 Chr.19: 6414108-6414108 on Build GRCh38
rs766744667 Chr.19: 6414212-6414212 on Build GRCh38
rs764927133 Chr.19: 6414234-6414234 on Build GRCh38
rs772339845 Chr.19: 6414228-6414228 on Build GRCh38
rs751723904 Chr.19: 6414180-6414180 on Build GRCh38
rs761283656 Chr.19: 6414071-6414071 on Build GRCh38

View Full Product Details