The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000367.3 | NP_000358.1 | 1 - 2 | 130 | 80 |
XM_011514839.2 | XP_011513141.1 | 1 - 2 | 88 | 80 | |
GenBank mRNA | BC005339.1 | - | 1 - 2 | 40 | 80 |
BC009596.1 | - | 1 - 2 | 39 | 80 | |
BI598325.1 | - | 1 - 2 | 115 | 80 | |
DB481604.1 | - | 1 - 2 | 134 | 80 | |
U12387.1 | - | 1 - 2 | 29 | 80 |