Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
79786
Gene Name
kelch like family member 36
Gene Aliases
C16orf44
Chromosome Location
Chr.16: 84648504 - 84667686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303451.1 NP_001290380.1 4 - 4 6392 91
NM_024731.3 NP_079007.2 5 - 5 6581 91
XM_005256149.2 XP_005256206.1 5 - 5 6795 91
XM_017023674.1 XP_016879163.1 5 - 5 6543 91
XM_017023675.1 XP_016879164.1 5 - 5 6563 91
GenBank mRNA AK056197.1 - 1 - 1 1575 91
AK056428.1 - 1 - 1 3379 91
AK074056.1 - 2 - 2 5508 91

View Full Product Details