The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031313.2 | NP_112603.2 | 2 - 3 | 237 | 124 |
GenBank mRNA | AK293017.1 | - | 2 - 3 | 235 | 124 |
BC014139.2 | - | 2 - 3 | 222 | 124 | |
J04948.1 | - | 2 - 3 | 236 | 124 | |
X53279.1 | - | 2 - 3 | 187 | 124 | |
X55958.1 | - | 2 - 3 | 214 | 124 |