Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96879765-96880038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPQ
Entrez Gene ID
10404
Gene Name
carboxypeptidase Q
Gene Aliases
LDP, PGCP
UniGene
Hs.156178
Gene Chromosome Location
Chr.8: 96645227-97143503 on Build GRCh38
Transcripts
NM_016134.3 XM_017012953.1 XM_017012954.1 XM_017012955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047879 c.694G>T p.A232S Chr.8: 96879850-96879850 on Build GRCh38
5007811 c.797C>A p.T266N Chr.8: 96879953-96879953 on Build GRCh38
3835190 c.707T>G p.V236G Chr.8: 96879863-96879863 on Build GRCh38
6634756 c.700A>G p.I234V Chr.8: 96879856-96879856 on Build GRCh38
3731755 c.705G>A p.T235T Chr.8: 96879861-96879861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770250110 Chr.8: 96879843-96879843 on Build GRCh38
rs763107508 Chr.8: 96879789-96879789 on Build GRCh38
rs748699520 Chr.8: 96879917-96879917 on Build GRCh38
rs771260509 Chr.8: 96879946-96879946 on Build GRCh38
rs774757454 Chr.8: 96879968-96879968 on Build GRCh38
rs753547633 Chr.8: 96879821-96879821 on Build GRCh38
rs756363859 Chr.8: 96879935-96879935 on Build GRCh38
rs753447649 Chr.8: 96879889-96879889 on Build GRCh38
rs564531366 Chr.8: 96879832-96879832 on Build GRCh38
rs759581285 Chr.8: 96879803-96879803 on Build GRCh38
rs774599864 Chr.8: 96879863-96879863 on Build GRCh38
rs112681566 Chr.8: 96879899-96879899 on Build GRCh38
rs141107654 Chr.8: 96879891-96879891 on Build GRCh38
rs368218633 Chr.8: 96879854-96879854 on Build GRCh38
rs375182115 Chr.8: 96879986-96879986 on Build GRCh38
rs372113350 Chr.8: 96879861-96879861 on Build GRCh38
rs977174043 Chr.8: 96879882-96879882 on Build GRCh38
rs151033390 Chr.8: 96879898-96879898 on Build GRCh38
rs781535494 Chr.8: 96879911-96879911 on Build GRCh38
rs61741750 Chr.8: 96879831-96879831 on Build GRCh38
rs771615579 Chr.8: 96879989-96879989 on Build GRCh38
rs560500593 Chr.8: 96879896-96879896 on Build GRCh38
rs769872674 Chr.8: 96879787-96879787 on Build GRCh38
rs374279890 Chr.8: 96879837-96879837 on Build GRCh38
rs199722608 Chr.8: 96879807-96879807 on Build GRCh38
rs778316900 Chr.8: 96879937-96879937 on Build GRCh38
rs140416726 Chr.8: 96879973-96879973 on Build GRCh38
rs751479155 Chr.8: 96879790-96879790 on Build GRCh38
rs755978642 Chr.8: 96879810-96879810 on Build GRCh38
rs778874767 Chr.8: 96879819-96879819 on Build GRCh38
rs749474745 Chr.8: 96879945-96879945 on Build GRCh38
rs201427381 Chr.8: 96879820-96879820 on Build GRCh38
rs993064538 Chr.8: 96879856-96879856 on Build GRCh38
rs778425620 Chr.8: 96879830-96879830 on Build GRCh38
rs201646330 Chr.8: 96879883-96879883 on Build GRCh38
rs776386723 Chr.8: 96880006-96880006 on Build GRCh38
rs969404373 Chr.8: 96879811-96879811 on Build GRCh38
rs149951331 Chr.8: 96880003-96880003 on Build GRCh38
rs775566173 Chr.8: 96879877-96879877 on Build GRCh38
rs775562223 Chr.8: 96879983-96879983 on Build GRCh38
rs935697062 Chr.8: 96879916-96879916 on Build GRCh38
rs772251568 Chr.8: 96879970-96879970 on Build GRCh38
rs200913578 Chr.8: 96879910-96879910 on Build GRCh38
rs575515198 Chr.8: 96879822-96879822 on Build GRCh38
rs745938647 Chr.8: 96879969-96879969 on Build GRCh38
rs147764297 Chr.8: 96879805-96879805 on Build GRCh38
rs139595101 Chr.8: 96879860-96879860 on Build GRCh38
rs763963550 Chr.8: 96879886-96879886 on Build GRCh38
rs955032340 Chr.8: 96879966-96879966 on Build GRCh38
rs972675589 Chr.8: 96879908-96879908 on Build GRCh38
rs759260946 Chr.8: 96879884-96879885 on Build GRCh38
rs757077941 Chr.8: 96879788-96879788 on Build GRCh38
rs578138575 Chr.8: 96879865-96879865 on Build GRCh38
rs778966186 Chr.8: 96879887-96879887 on Build GRCh38

View Full Product Details