The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178525.4 | NP_848620.3 | 1 - 1 | 259 | 124 |
GenBank mRNA | AK292258.1 | - | 1 - 1 | 207 | 124 |
AY248901.1 | - | 1 - 1 | 134 | 124 | |
BC024028.2 | - | 1 - 1 | 207 | 124 | |
BC045752.1 | - | 1 - 1 | 251 | 124 | |
HY011370.1 | - | 1 - 1 | 259 | 124 | |
HY042660.1 | - | 1 - 1 | 225 | 124 |