Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC100132686
- Entrez Gene ID
-
100132686
- Gene Name
- uncharacterized LOC100132686
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 112260265 - 112261396 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_133566.1 |
|
1 - 1 |
551 |
101 |
GenBank mRNA |
BC020894.1 |
- |
1 - 1 |
551 |
101 |
- Gene Symbol
- PLET1
- Entrez Gene ID
-
349633
- Gene Name
- placenta expressed transcript 1
- Gene Aliases
-
C11orf34
- Chromosome Location
-
Chr.11: 112248153 - 112260860 on Build GRCh38
View Full Product Details