The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002834.3 | NP_002825.3 | 16 - 16 | 4888 | 88 |
XM_006719526.2 | XP_006719589.1 | 16 - 16 | 4678 | 88 | |
XM_011538613.2 | XP_011536915.1 | 16 - 16 | 4714 | 88 | |
XM_017019722.1 | XP_016875211.1 | 16 - 16 | 4705 | 88 | |
GenBank mRNA | BC007869.2 | - | 1 - 1 | 377 | 88 |
BC025181.1 | - | 1 - 1 | 453 | 88 | |
BC030949.1 | - | 1 - 1 | 397 | 88 | |
BX537632.1 | - | 1 - 1 | 1977 | 88 |