Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215194772-215195033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK2
Entrez Gene ID
3776
Gene Name
potassium two pore domain channel subfamily K member 2
Gene Aliases
K2p2.1, TPKC1, TREK, TREK-1, TREK1, hTREK-1c, hTREK-1e
UniGene
Hs.497745
Gene Chromosome Location
Chr.1: 215002920-215237093 on Build GRCh38
Transcripts
NM_001017424.2 NM_001017425.2 NM_014217.3 XM_011509522.2 XM_011509523.1 XM_011509524.2 XM_017001248.1 XM_017001249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211574 c.820G>A p.V274M Chr.1: 215194994-215194994 on Build GRCh38
6483068 c.881T>A p.L294H Chr.1: 215195010-215195010 on Build GRCh38
6124472 c.897C>A p.Y299* Chr.1: 215195026-215195026 on Build GRCh38
4957400 c.827G>A p.G276E Chr.1: 215194956-215194956 on Build GRCh38
5161315 c.819C>T p.S273S Chr.1: 215194960-215194960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374646942 Chr.1: 215194976-215194976 on Build GRCh38
rs565618538 Chr.1: 215194993-215194993 on Build GRCh38
rs769674825 Chr.1: 215194983-215194983 on Build GRCh38
rs961009632 Chr.1: 215194879-215194879 on Build GRCh38
rs773704506 Chr.1: 215194918-215194918 on Build GRCh38
rs763152132 Chr.1: 215194950-215194950 on Build GRCh38
rs543257957 Chr.1: 215194901-215194901 on Build GRCh38
rs771316818 Chr.1: 215194922-215194922 on Build GRCh38
rs908595713 Chr.1: 215194890-215194890 on Build GRCh38
rs940529033 Chr.1: 215194899-215194899 on Build GRCh38
rs534747722 Chr.1: 215194827-215194827 on Build GRCh38
rs745349220 Chr.1: 215194902-215194902 on Build GRCh38
rs755608318 Chr.1: 215194906-215194906 on Build GRCh38
rs371312018 Chr.1: 215194964-215194964 on Build GRCh38
rs981677411 Chr.1: 215194806-215194806 on Build GRCh38
rs768480592 Chr.1: 215194917-215194917 on Build GRCh38
rs775563802 Chr.1: 215194945-215194945 on Build GRCh38
rs928895239 Chr.1: 215194810-215194810 on Build GRCh38
rs767240406 Chr.1: 215194966-215194966 on Build GRCh38
rs776868050 Chr.1: 215195012-215195012 on Build GRCh38
rs972405923 Chr.1: 215194880-215194880 on Build GRCh38
rs747823400 Chr.1: 215194996-215194996 on Build GRCh38
rs750307755 Chr.1: 215194968-215194968 on Build GRCh38
rs764366307 Chr.1: 215194956-215194956 on Build GRCh38
rs765297963 Chr.1: 215194942-215194942 on Build GRCh38
rs779432870 Chr.1: 215194914-215194914 on Build GRCh38
rs577641352 Chr.1: 215194894-215194894 on Build GRCh38
rs749010897 Chr.1: 215194959-215194959 on Build GRCh38
rs748891121 Chr.1: 215194916-215194916 on Build GRCh38
rs149941939 Chr.1: 215194960-215194960 on Build GRCh38
rs573638555 Chr.1: 215194969-215194969 on Build GRCh38
rs747415049 Chr.1: 215194921-215194921 on Build GRCh38
rs752327424 Chr.1: 215194989-215194989 on Build GRCh38
rs759994903 Chr.1: 215194940-215194940 on Build GRCh38
rs771290454 Chr.1: 215195004-215195004 on Build GRCh38
rs557586161 Chr.1: 215194828-215194828 on Build GRCh38

View Full Product Details