Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEFTY2
Entrez Gene ID
7044
Gene Name
left-right determination factor 2
Gene Aliases
EBAF, LEFTA, LEFTYA, TGFB4
Chromosome Location
Chr.1: 225936598 - 225941383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172425.1 NP_001165896.1 5 - 5 1991 102
NM_003240.3 NP_003231.2 4 - 4 2093 102
GenBank mRNA AF081513.1 - 4 - 4 1918 102
AK027520.1 - 4 - 4 1930 102
AK075344.1 - 4 - 4 1930 102
AK094077.1 - - 1696 102
BC035718.1 - 4 - 4 1918 102
BC065929.1 - 2 - 2 1715 102
U81523.1 - 4 - 4 1875 102

View Full Product Details