The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021979.3 | NP_068814.2 | 1 - 1 | 2521 | 152 |
GenBank mRNA | BC001752.1 | - | 2 - 2 | 2215 | 152 |
BC036107.1 | - | 1 - 1 | 2249 | 152 |