Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10290895-10291166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM5
Entrez Gene ID
7087
Gene Name
intercellular adhesion molecule 5
Gene Aliases
TLCN, TLN
UniGene
Hs.465862
Gene Chromosome Location
Chr.19: 10289352-10296778 on Build GRCh38
Transcripts
NM_003259.3 XM_011528229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4073782 c.84G>A p.A28A Chr.19: 10291073-10291073 on Build GRCh38
4073783 c.128C>T p.A43V Chr.19: 10291117-10291117 on Build GRCh38
1241288 c.122G>A p.R41H Chr.19: 10291111-10291111 on Build GRCh38
6539030 c.149C>G p.S50W Chr.19: 10291138-10291138 on Build GRCh38
1680636 c.100T>G p.F34V Chr.19: 10291089-10291089 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs976570222 Chr.19: 10290933-10290933 on Build GRCh38
rs986282463 Chr.19: 10291019-10291019 on Build GRCh38
rs2075742 Chr.19: 10290931-10290931 on Build GRCh38
rs370741150 Chr.19: 10291024-10291024 on Build GRCh38
rs1021038215 Chr.19: 10290959-10290959 on Build GRCh38
rs371662961 Chr.19: 10291063-10291063 on Build GRCh38
rs1049631085 Chr.19: 10291111-10291111 on Build GRCh38
rs781488060 Chr.19: 10291044-10291044 on Build GRCh38
rs771041441 Chr.19: 10291058-10291058 on Build GRCh38
rs770220683 Chr.19: 10291046-10291046 on Build GRCh38
rs1013239814 Chr.19: 10290936-10290936 on Build GRCh38
rs80066865 Chr.19: 10290998-10290998 on Build GRCh38
rs896310267 Chr.19: 10291039-10291039 on Build GRCh38
rs745487938 Chr.19: 10291032-10291032 on Build GRCh38
rs573140592 Chr.19: 10291017-10291017 on Build GRCh38
rs1036307052 Chr.19: 10291038-10291038 on Build GRCh38
rs568068970 Chr.19: 10291072-10291072 on Build GRCh38
rs763846097 Chr.19: 10291100-10291100 on Build GRCh38
rs773785177 Chr.19: 10291053-10291053 on Build GRCh38
rs376570687 Chr.19: 10291079-10291079 on Build GRCh38
rs775389277 Chr.19: 10291086-10291086 on Build GRCh38
rs540436743 Chr.19: 10291021-10291021 on Build GRCh38
rs760572744 Chr.19: 10291096-10291096 on Build GRCh38
rs929295595 Chr.19: 10291082-10291082 on Build GRCh38
rs756777035 Chr.19: 10291029-10291029 on Build GRCh38
rs931933556 Chr.19: 10291012-10291012 on Build GRCh38
rs568816242 Chr.19: 10290957-10290957 on Build GRCh38
rs369518200 Chr.19: 10291142-10291142 on Build GRCh38
rs555553140 Chr.19: 10291031-10291031 on Build GRCh38
rs749820628 Chr.19: 10291055-10291055 on Build GRCh38
rs759815089 Chr.19: 10291073-10291073 on Build GRCh38
rs539376521 Chr.19: 10290967-10290967 on Build GRCh38
rs2569698 Chr.19: 10290964-10290964 on Build GRCh38
rs748544440 Chr.19: 10291045-10291045 on Build GRCh38
rs758095126 Chr.19: 10291043-10291043 on Build GRCh38

View Full Product Details