Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABT1
Entrez Gene ID
29777
Gene Name
activator of basal transcription 1
Gene Aliases
hABT1
Chromosome Location
Chr.6: 26596943 - 26600050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013375.3 NP_037507.1 3 - 3 1040 142
GenBank mRNA AB027258.1 - 3 - 3 1020 142
BC048812.1 - 3 - 3 1038 142
BC066313.1 - 3 - 3 1030 142

View Full Product Details