Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21807293-21807552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC9
Entrez Gene ID
10060
Gene Name
ATP binding cassette subfamily C member 9
Gene Aliases
ABC37, ATFB12, CANTU, CMD1O, SUR2
UniGene
Hs.732701
Gene Chromosome Location
Chr.12: 21797389-21941869 on Build GRCh38
Transcripts
NM_005691.3 NM_020297.3 XM_005253284.3 XM_005253286.3 XM_005253287.4 XM_005253288.3 XM_005253289.3 XM_005253290.3 XM_006719025.3 XM_011520545.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1360701 c.4424C>A p.A1475E Chr.12: 21807371-21807371 on Build GRCh38
3459253 c.4371T>A p.F1457L Chr.12: 21807424-21807424 on Build GRCh38
937992 c.4319C>T p.A1440V Chr.12: 21807476-21807476 on Build GRCh38
3459255 c.4338G>A p.G1446G Chr.12: 21807457-21807457 on Build GRCh38
7133495 c.4411A>T p.I1471F Chr.12: 21807384-21807384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368079660 Chr.12: 21807358-21807358 on Build GRCh38
rs371653292 Chr.12: 21807403-21807403 on Build GRCh38
rs530506599 Chr.12: 21807406-21807406 on Build GRCh38
rs867034453 Chr.12: 21807401-21807401 on Build GRCh38
rs766413117 Chr.12: 21807439-21807440 on Build GRCh38
rs766276761 Chr.12: 21807511-21807511 on Build GRCh38
rs931320329 Chr.12: 21807390-21807390 on Build GRCh38
rs1012250973 Chr.12: 21807510-21807510 on Build GRCh38
rs771226136 Chr.12: 21807442-21807442 on Build GRCh38
rs367697857 Chr.12: 21807498-21807498 on Build GRCh38
rs767586816 Chr.12: 21807476-21807476 on Build GRCh38
rs889569693 Chr.12: 21807483-21807483 on Build GRCh38
rs554735392 Chr.12: 21807482-21807482 on Build GRCh38
rs1005698399 Chr.12: 21807508-21807508 on Build GRCh38
rs763562261 Chr.12: 21807470-21807470 on Build GRCh38
rs377289768 Chr.12: 21807475-21807475 on Build GRCh38
rs780440555 Chr.12: 21807318-21807318 on Build GRCh38
rs1050347970 Chr.12: 21807458-21807458 on Build GRCh38
rs146782703 Chr.12: 21807451-21807451 on Build GRCh38
rs895232384 Chr.12: 21807467-21807468 on Build GRCh38
rs750331807 Chr.12: 21807490-21807490 on Build GRCh38
rs140448278 Chr.12: 21807473-21807473 on Build GRCh38
rs747395248 Chr.12: 21807441-21807441 on Build GRCh38
rs397517190 Chr.12: 21807443-21807443 on Build GRCh38
rs886049168 Chr.12: 21807493-21807493 on Build GRCh38
rs749457961 Chr.12: 21807328-21807328 on Build GRCh38
rs755085104 Chr.12: 21807355-21807355 on Build GRCh38
rs768813655 Chr.12: 21807509-21807509 on Build GRCh38
rs727504785 Chr.12: 21807430-21807430 on Build GRCh38

View Full Product Details