Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37966237-37966510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRB3
Entrez Gene ID
155
Gene Name
adrenoceptor beta 3
Gene Aliases
BETA3AR
UniGene
Hs.2549
Gene Chromosome Location
Chr.8: 37962995-37966666 on Build GRCh38
Transcripts
NM_000025.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834645 c.186C>G p.I62M Chr.8: 37966284-37966284 on Build GRCh38
5680974 c.213G>T p.M71I Chr.8: 37966257-37966257 on Build GRCh38
4985656 c.190T>C p.W64R Chr.8: 37966280-37966280 on Build GRCh38
3834644 c.198G>A p.P66P Chr.8: 37966272-37966272 on Build GRCh38
5358858 c.212T>C p.M71T Chr.8: 37966258-37966258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549662633 Chr.8: 37966273-37966273 on Build GRCh38
rs377609934 Chr.8: 37966374-37966374 on Build GRCh38
rs202143668 Chr.8: 37966416-37966416 on Build GRCh38
rs953228215 Chr.8: 37966418-37966418 on Build GRCh38
rs760780325 Chr.8: 37966461-37966461 on Build GRCh38
rs202110322 Chr.8: 37966462-37966462 on Build GRCh38
rs201243226 Chr.8: 37966452-37966452 on Build GRCh38
rs201212770 Chr.8: 37966425-37966425 on Build GRCh38
rs749594748 Chr.8: 37966430-37966430 on Build GRCh38
rs759812310 Chr.8: 37966449-37966449 on Build GRCh38
rs949039693 Chr.8: 37966376-37966376 on Build GRCh38
rs771352006 Chr.8: 37966431-37966431 on Build GRCh38
rs200142610 Chr.8: 37966421-37966421 on Build GRCh38
rs201549798 Chr.8: 37966321-37966321 on Build GRCh38
rs199996398 Chr.8: 37966346-37966346 on Build GRCh38
rs758260065 Chr.8: 37966389-37966389 on Build GRCh38
rs941748993 Chr.8: 37966347-37966347 on Build GRCh38
rs965382092 Chr.8: 37966286-37966286 on Build GRCh38
rs772084969 Chr.8: 37966454-37966454 on Build GRCh38
rs200305091 Chr.8: 37966442-37966442 on Build GRCh38
rs757169350 Chr.8: 37966359-37966359 on Build GRCh38
rs762614429 Chr.8: 37966474-37966474 on Build GRCh38
rs1009625001 Chr.8: 37966441-37966441 on Build GRCh38
rs370860214 Chr.8: 37966426-37966426 on Build GRCh38
rs766199434 Chr.8: 37966475-37966475 on Build GRCh38
rs28364013 Chr.8: 37966358-37966358 on Build GRCh38
rs781058113 Chr.8: 37966488-37966488 on Build GRCh38
rs909045042 Chr.8: 37966318-37966318 on Build GRCh38
rs757977808 Chr.8: 37966282-37966282 on Build GRCh38
rs368109921 Chr.8: 37966356-37966356 on Build GRCh38
rs779522839 Chr.8: 37966390-37966390 on Build GRCh38
rs201031255 Chr.8: 37966413-37966413 on Build GRCh38
rs774686772 Chr.8: 37966344-37966344 on Build GRCh38
rs200478962 Chr.8: 37966473-37966473 on Build GRCh38
rs201389673 Chr.8: 37966292-37966292 on Build GRCh38
rs200514376 Chr.8: 37966369-37966369 on Build GRCh38
rs4994 Chr.8: 37966280-37966280 on Build GRCh38
rs199751425 Chr.8: 37966296-37966296 on Build GRCh38
rs754665596 Chr.8: 37966481-37966481 on Build GRCh38
rs765221550 Chr.8: 37966365-37966365 on Build GRCh38
rs757009547 Chr.8: 37966263-37966263 on Build GRCh38
rs772373419 Chr.8: 37966342-37966342 on Build GRCh38
rs919039334 Chr.8: 37966360-37966360 on Build GRCh38
rs773698704 Chr.8: 37966330-37966330 on Build GRCh38
rs765135471 Chr.8: 37966468-37966468 on Build GRCh38
rs767619305 Chr.8: 37966349-37966349 on Build GRCh38
rs965476467 Chr.8: 37966471-37966471 on Build GRCh38
rs202130418 Chr.8: 37966435-37966435 on Build GRCh38
rs201607471 Chr.8: 37966397-37966397 on Build GRCh38
rs953458416 Chr.8: 37966297-37966297 on Build GRCh38
rs892525758 Chr.8: 37966329-37966329 on Build GRCh38
rs751212170 Chr.8: 37966358-37966358 on Build GRCh38
rs367948517 Chr.8: 37966422-37966422 on Build GRCh38
rs200051861 Chr.8: 37966314-37966314 on Build GRCh38
rs751114839 Chr.8: 37966477-37966477 on Build GRCh38
rs200049119 Chr.8: 37966412-37966412 on Build GRCh38
rs201940081 Chr.8: 37966458-37966458 on Build GRCh38
rs866356810 Chr.8: 37966467-37966467 on Build GRCh38
rs779794484 Chr.8: 37966284-37966284 on Build GRCh38
rs763443477 Chr.8: 37966335-37966335 on Build GRCh38
rs147723870 Chr.8: 37966317-37966317 on Build GRCh38

View Full Product Details