Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11298078-11298323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELX
Entrez Gene ID
265
Gene Name
amelogenin, X-linked
Gene Aliases
AI1E, AIH1, ALGN, AMG, AMGL, AMGX
UniGene
Hs.654436
Gene Chromosome Location
Chr.X: 11293413-11300761 on Build GRCh38
Transcripts
NM_001142.2 NM_182680.1 NM_182681.1 XM_017029404.1 NM_001287242.1 NM_006125.2 NM_013427.2 NR_109776.1

Gene Symbol
ARHGAP6
Entrez Gene ID
395
Gene Name
Rho GTPase activating protein 6
Gene Aliases
RHOGAP6, RHOGAPX-1
UniGene
Hs.435291
Gene Chromosome Location
Chr.X: 11137543-11665701 on Build GRCh38
Transcripts
NM_001142.2 NM_182680.1 NM_182681.1 XM_017029404.1 NM_001287242.1 NM_006125.2 NM_013427.2 NR_109776.1

SNP Detail

SNP ID Chromosome Location
rs199717275 Chr.X: 11298181-11298181 on Build GRCh38
rs746714110 Chr.X: 11298125-11298125 on Build GRCh38
rs867528957 Chr.X: 11298137-11298137 on Build GRCh38
rs765738846 Chr.X: 11298256-11298256 on Build GRCh38
rs144955572 Chr.X: 11298276-11298276 on Build GRCh38
rs373696473 Chr.X: 11298162-11298162 on Build GRCh38
rs762554859 Chr.X: 11298211-11298211 on Build GRCh38
rs773161383 Chr.X: 11298170-11298170 on Build GRCh38
rs149068672 Chr.X: 11298277-11298277 on Build GRCh38
rs431825176 Chr.X: 11298262-11298262 on Build GRCh38
rs767685604 Chr.X: 11298184-11298184 on Build GRCh38
rs776437048 Chr.X: 11298196-11298197 on Build GRCh38
rs375383821 Chr.X: 11298287-11298287 on Build GRCh38
rs765286677 Chr.X: 11298166-11298166 on Build GRCh38
rs779323924 Chr.X: 11298235-11298235 on Build GRCh38
rs431825177 Chr.X: 11298264-11298264 on Build GRCh38
rs994543951 Chr.X: 11298283-11298283 on Build GRCh38
rs768417162 Chr.X: 11298149-11298149 on Build GRCh38
rs996514040 Chr.X: 11298115-11298115 on Build GRCh38
rs1035216478 Chr.X: 11298257-11298257 on Build GRCh38
rs200163085 Chr.X: 11298155-11298155 on Build GRCh38
rs201515468 Chr.X: 11298154-11298154 on Build GRCh38
rs961496168 Chr.X: 11298228-11298228 on Build GRCh38
rs104894733 Chr.X: 11298243-11298243 on Build GRCh38
rs765441386 Chr.X: 11298180-11298180 on Build GRCh38
rs752708064 Chr.X: 11298187-11298187 on Build GRCh38
rs754292311 Chr.X: 11298207-11298207 on Build GRCh38
rs431825178 Chr.X: 11298265-11298265 on Build GRCh38
rs387906487 Chr.X: 11298246-11298246 on Build GRCh38
rs764237259 Chr.X: 11298219-11298219 on Build GRCh38
rs754218426 Chr.X: 11298223-11298223 on Build GRCh38
rs759477208 Chr.X: 11298221-11298221 on Build GRCh38
rs758813809 Chr.X: 11298272-11298272 on Build GRCh38

View Full Product Details