Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15379939-15380188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
UniGene
Hs.399800
Gene Chromosome Location
Chr.19: 15380048-15419121 on Build GRCh38
Transcripts
NM_001291478.1 NM_014371.3 NR_111971.1 XM_005259854.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7321642 c.1890C>T p.I630I Chr.19: 15380173-15380173 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897334178 Chr.19: 15380057-15380057 on Build GRCh38
rs1016408150 Chr.19: 15380116-15380116 on Build GRCh38
rs1010166137 Chr.19: 15380098-15380098 on Build GRCh38
rs1050858618 Chr.19: 15380066-15380066 on Build GRCh38
rs1006540139 Chr.19: 15380082-15380082 on Build GRCh38
rs941632806 Chr.19: 15380045-15380045 on Build GRCh38
rs573112236 Chr.19: 15380055-15380055 on Build GRCh38
rs759414231 Chr.19: 15380119-15380119 on Build GRCh38
rs752985263 Chr.19: 15380138-15380138 on Build GRCh38
rs998351048 Chr.19: 15380063-15380064 on Build GRCh38
rs941580518 Chr.19: 15380134-15380134 on Build GRCh38
rs553187478 Chr.19: 15380006-15380006 on Build GRCh38
rs11879108 Chr.19: 15380012-15380012 on Build GRCh38
rs537820331 Chr.19: 15380006-15380007 on Build GRCh38
rs959743344 Chr.19: 15380126-15380126 on Build GRCh38
rs1040988962 Chr.19: 15379987-15379987 on Build GRCh38
rs545940271 Chr.19: 15379959-15379959 on Build GRCh38
rs758408137 Chr.19: 15380139-15380139 on Build GRCh38
rs542140175 Chr.19: 15380103-15380103 on Build GRCh38
rs893030892 Chr.19: 15380089-15380089 on Build GRCh38
rs866372438 Chr.19: 15380053-15380053 on Build GRCh38
rs919545150 Chr.19: 15379988-15380004 on Build GRCh38
rs959542098 Chr.19: 15380132-15380132 on Build GRCh38
rs928076681 Chr.19: 15379988-15379988 on Build GRCh38
rs974150009 Chr.19: 15379989-15379989 on Build GRCh38
rs759328757 Chr.19: 15380164-15380164 on Build GRCh38
rs760793998 Chr.19: 15379977-15379977 on Build GRCh38
rs1037131029 Chr.19: 15380048-15380048 on Build GRCh38
rs776453029 Chr.19: 15380148-15380159 on Build GRCh38
rs1035096600 Chr.19: 15380125-15380125 on Build GRCh38
rs765223433 Chr.19: 15380149-15380149 on Build GRCh38
rs886512700 Chr.19: 15380096-15380096 on Build GRCh38
rs972959396 Chr.19: 15380140-15380140 on Build GRCh38
rs938018105 Chr.19: 15379995-15379995 on Build GRCh38
rs997512326 Chr.19: 15380072-15380072 on Build GRCh38
rs918875989 Chr.19: 15380161-15380161 on Build GRCh38
rs562017892 Chr.19: 15380120-15380120 on Build GRCh38
rs752012911 Chr.19: 15380167-15380167 on Build GRCh38
rs974052934 Chr.19: 15379984-15380000 on Build GRCh38
rs889564066 Chr.19: 15380076-15380076 on Build GRCh38
rs911077899 Chr.19: 15379968-15379968 on Build GRCh38
rs1006729917 Chr.19: 15380113-15380113 on Build GRCh38
rs990810900 Chr.19: 15380130-15380130 on Build GRCh38
rs764173803 Chr.19: 15380159-15380159 on Build GRCh38
rs910116086 Chr.19: 15380007-15380011 on Build GRCh38
rs548504573 Chr.19: 15380133-15380133 on Build GRCh38
rs971217449 Chr.19: 15380107-15380107 on Build GRCh38
rs766144118 Chr.19: 15380086-15380086 on Build GRCh38

View Full Product Details