Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15678194-15678451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
UniGene
Hs.335239
Gene Chromosome Location
Chr.3: 15667236-15859814 on Build GRCh38
Transcripts
NM_001195098.1 NM_001195099.1 NM_015199.3 XM_005264996.3 XM_011533540.2 XM_011533541.2 XM_011533542.2 XM_011533543.2 XM_011533545.2 XM_011533546.2 XM_011533547.2 XM_017006026.1 XM_017006027.1 XM_017006029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584064 c.955G>C p.V319L Chr.3: 15678272-15678272 on Build GRCh38
3589346 c.883C>T p.H295Y Chr.3: 15678344-15678344 on Build GRCh38
6853761 c.2633A>G p.H878R Chr.3: 15678292-15678292 on Build GRCh38
4423032 c.962C>G p.S321C Chr.3: 15678265-15678265 on Build GRCh38
367324 c.983T>C p.M328T Chr.3: 15678244-15678244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200889611 Chr.3: 15678315-15678315 on Build GRCh38
rs756030468 Chr.3: 15678401-15678401 on Build GRCh38
rs141022482 Chr.3: 15678421-15678421 on Build GRCh38
rs778561547 Chr.3: 15678317-15678317 on Build GRCh38
rs761189148 Chr.3: 15678248-15678248 on Build GRCh38
rs1055405609 Chr.3: 15678395-15678395 on Build GRCh38
rs756089133 Chr.3: 15678295-15678295 on Build GRCh38
rs750618889 Chr.3: 15678260-15678260 on Build GRCh38
rs777137741 Chr.3: 15678347-15678347 on Build GRCh38
rs753830845 Chr.3: 15678404-15678404 on Build GRCh38
rs752589114 Chr.3: 15678283-15678283 on Build GRCh38
rs994313540 Chr.3: 15678265-15678265 on Build GRCh38
rs929821181 Chr.3: 15678308-15678308 on Build GRCh38
rs369691636 Chr.3: 15678311-15678311 on Build GRCh38
rs750611281 Chr.3: 15678358-15678358 on Build GRCh38
rs373040079 Chr.3: 15678371-15678371 on Build GRCh38
rs780872483 Chr.3: 15678281-15678281 on Build GRCh38
rs963976883 Chr.3: 15678398-15678398 on Build GRCh38
rs754766441 Chr.3: 15678277-15678277 on Build GRCh38
rs777925632 Chr.3: 15678297-15678297 on Build GRCh38
rs761064604 Chr.3: 15678337-15678337 on Build GRCh38
rs753880632 Chr.3: 15678254-15678254 on Build GRCh38
rs769141016 Chr.3: 15678344-15678344 on Build GRCh38
rs764631967 Chr.3: 15678252-15678252 on Build GRCh38
rs776021988 Chr.3: 15678241-15678241 on Build GRCh38
rs752537862 Chr.3: 15678397-15678397 on Build GRCh38
rs372111227 Chr.3: 15678259-15678259 on Build GRCh38
rs775806991 Chr.3: 15678336-15678336 on Build GRCh38
rs745588421 Chr.3: 15678318-15678318 on Build GRCh38
rs745460320 Chr.3: 15678257-15678257 on Build GRCh38
rs764063969 Chr.3: 15678403-15678403 on Build GRCh38
rs766665773 Chr.3: 15678377-15678377 on Build GRCh38
rs765267091 Chr.3: 15678353-15678353 on Build GRCh38
rs368204050 Chr.3: 15678240-15678240 on Build GRCh38
rs551351650 Chr.3: 15678423-15678423 on Build GRCh38
rs772429036 Chr.3: 15678322-15678322 on Build GRCh38
rs761818731 Chr.3: 15678350-15678350 on Build GRCh38

View Full Product Details