Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241559299-241559544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOK
Entrez Gene ID
666
Gene Name
BOK, BCL2 family apoptosis regulator
Gene Aliases
BCL2L9, BOKL
UniGene
Hs.293753
Gene Chromosome Location
Chr.2: 241558731-241574138 on Build GRCh38
Transcripts
NM_032515.4 XM_011511696.2 XM_011511697.2 XM_017004775.1 XM_017004776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378523 c.35C>T p.A12V Chr.2: 241559518-241559518 on Build GRCh38
4802740 c.24G>T p.S8S Chr.2: 241559507-241559507 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959835553 Chr.2: 241559455-241559455 on Build GRCh38
rs893168949 Chr.2: 241559404-241559404 on Build GRCh38
rs201959873 Chr.2: 241559526-241559526 on Build GRCh38
rs771747908 Chr.2: 241559470-241559470 on Build GRCh38
rs947077971 Chr.2: 241559518-241559518 on Build GRCh38
rs772965442 Chr.2: 241559485-241559485 on Build GRCh38
rs569000629 Chr.2: 241559468-241559468 on Build GRCh38
rs760213746 Chr.2: 241559493-241559493 on Build GRCh38
rs1028970710 Chr.2: 241559376-241559376 on Build GRCh38
rs1040869028 Chr.2: 241559384-241559384 on Build GRCh38
rs868291562 Chr.2: 241559514-241559514 on Build GRCh38
rs562127739 Chr.2: 241559397-241559397 on Build GRCh38
rs765974657 Chr.2: 241559506-241559506 on Build GRCh38
rs533395543 Chr.2: 241559516-241559516 on Build GRCh38
rs1053525070 Chr.2: 241559395-241559395 on Build GRCh38
rs965273138 Chr.2: 241559520-241559520 on Build GRCh38
rs985429881 Chr.2: 241559353-241559353 on Build GRCh38
rs911043103 Chr.2: 241559375-241559375 on Build GRCh38
rs982176509 Chr.2: 241559418-241559418 on Build GRCh38
rs758981632 Chr.2: 241559515-241559515 on Build GRCh38
rs911108077 Chr.2: 241559503-241559503 on Build GRCh38
rs952735793 Chr.2: 241559352-241559352 on Build GRCh38
rs923324405 Chr.2: 241559389-241559389 on Build GRCh38
rs774361001 Chr.2: 241559466-241559466 on Build GRCh38
rs972777664 Chr.2: 241559321-241559321 on Build GRCh38
rs919605839 Chr.2: 241559347-241559347 on Build GRCh38
rs1015600848 Chr.2: 241559402-241559402 on Build GRCh38
rs776139987 Chr.2: 241559507-241559507 on Build GRCh38
rs1005925916 Chr.2: 241559391-241559391 on Build GRCh38
rs550663419 Chr.2: 241559427-241559427 on Build GRCh38
rs770111647 Chr.2: 241559502-241559503 on Build GRCh38
rs901337719 Chr.2: 241559358-241559358 on Build GRCh38
rs1003016329 Chr.2: 241559439-241559439 on Build GRCh38
rs993971694 Chr.2: 241559462-241559462 on Build GRCh38
rs1028549180 Chr.2: 241559429-241559429 on Build GRCh38
rs997132679 Chr.2: 241559373-241559373 on Build GRCh38
rs888460701 Chr.2: 241559387-241559387 on Build GRCh38
rs768725431 Chr.2: 241559456-241559456 on Build GRCh38
rs1012227592 Chr.2: 241559406-241559406 on Build GRCh38
rs529242810 Chr.2: 241559407-241559407 on Build GRCh38

View Full Product Details